Skip to Content
Merck
All Photos(2)

Documents

H3264

Sigma-Aldrich

n-Heptyl β-D-thioglucopyranoside

≥99% (GC)

Synonym(s):

Heptyl thioglucoside, Heptyl-β-D-1-thioglucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H26O5S
CAS Number:
Molecular Weight:
294.41
Beilstein:
6057483
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Quality Level

Assay

≥99% (GC)

form

powder

mol wt

294.41 g/mol

technique(s)

gas chromatography (GC): suitable

color

white

CMC

30

mp

98  °C ((208 °F ))

solubility

methanol: 100 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CCCCCCCS[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C13H26O5S/c1-2-3-4-5-6-7-19-13-12(17)11(16)10(15)9(8-14)18-13/h9-17H,2-8H2,1H3/t9-,10-,11+,12-,13+/m1/s1

InChI key

HPEGNLMTTNTJSP-LBELIVKGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful non-ionic detergent for solubilization and reconstitution of membrane proteins.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Sone
Journal of biochemistry, 100(6), 1465-1470 (1986-12-01)
It is possible to prepare liposomal vesicles by solubilization of total bacterial membranes with n-heptyl beta-D-thioglucoside followed by reconstitution into proteoliposomes by a freeze-thaw-sonication procedure with soybean phospholipids. The resulting proteoliposomes from total membrane fraction of sufficiently aerated cells of
T Ohyashiki et al.
Biochimica et biophysica acta, 902(1), 46-52 (1987-08-07)
The brush-border membrane from the porcine small intestine possesses Ca2+-dependent ATPase activity. The Ca2+ stimulation of ATP hydrolysis by the membranes is biphasic with a high affinity (Km = 0.38 microM) and a low affinity (Km = 98.3 microM). Treatment
K Nagai et al.
Journal of biochemistry, 101(5), 1115-1127 (1987-05-01)
Some properties of the enzyme activity that catalyzes the transfer of N-acetylgalactosamine from UDP-N-acetylgalactosamine to exogenous lactosylceramide-II3-sulfate (SM3) and N-acetylneuraminosyllactosylceramide (GM3) were studied using the enzyme preparation solubilized from the 100,000 X g pellet of 6-day-old rat brain. The products
T Shimamoto et al.
Journal of biochemistry, 97(6), 1807-1810 (1985-06-01)
A new nonionic detergent, heptylthioglucoside, was synthesized and found to be more soluble than octylthioglucoside in water at low temperatures. Use of this detergent for solubilization and reconstitution of membrane proteins of Escherichia coli was examined. Heptylthioglucoside was as effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service