Skip to Content
Merck
All Photos(1)

Documents

G10105

Sigma-Aldrich

18β-Glycyrrhetinic acid

97%

Synonym(s):

3β-Hydroxy-11-oxo-18β,20β-olean-12-en-29-oic acid, Enoxolone, Glycyrrhetin, Subglycyrrhelinic acid, Uralenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H46O4
CAS Number:
Molecular Weight:
470.68
Beilstein:
2229654
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]22/D +170.0°, c = 1 in chloroform

mp

292-295 °C (lit.)

SMILES string

CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(O)=O

InChI

1S/C30H46O4/c1-25(2)21-8-11-30(7)23(28(21,5)10-9-22(25)32)20(31)16-18-19-17-27(4,24(33)34)13-12-26(19,3)14-15-29(18,30)6/h16,19,21-23,32H,8-15,17H2,1-7H3,(H,33,34)/t19-,21-,22-,23+,26+,27-,28-,29+,30+/m0/s1

InChI key

MPDGHEJMBKOTSU-YKLVYJNSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

18β-Glycyrrhetinic acid is a pentacyclic triterpenoid found in the Glycyrrhiza glabra L.(liquorice) roots. It is the key metabolite of glycyrrhizin and glycyrrhizic acid.

Application

The product may be used as a starting material to prepare 18β-glycyrrhetinic acid derivatives, which show anti-inflammatory and antioxidant properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Stanetty et al.
Bioorganic & medicinal chemistry, 18(21), 7522-7541 (2010-09-21)
Glycyrrhetinic acid, the metabolite of the natural product glycyrrhizin, is a well known nonselective inhibitor of 11β-hydroxysteroid dehydrogenase (11β-HSD) type 1 and type 2. Whereas inhibition of 11β-HSD1 is currently under consideration for treatment of metabolic diseases, such as obesity
Igor Beseda et al.
Bioorganic & medicinal chemistry, 18(1), 433-454 (2009-11-17)
The effect of glycyrrhetinic acid (GA) and GA-derivatives towards 11beta-hydroxysteroid dehydrogenase (11beta-HSD) was investigated. Novel compounds with modifications at positions C-3, C-11 and C-29 of the GA skeleton were prepared. Single crystal X-ray diffraction data of selected substances are reported
Rawindra Gaware et al.
Bioorganic & medicinal chemistry, 19(6), 1866-1880 (2011-03-08)
Glycyrrhetinic acid, the metabolite of the natural product glycyrrhizin, is a well known nonselective inhibitor of 11β-hydroxysteroid dehydrogenase (11β-HSD) type 1 and type 2. Whereas inhibition of 11β-HSD1 is currently under consideration for treatment of metabolic diseases, such as obesity
A robust screening method for dietary agents that activate tumour-suppressor microRNAs.
Hagiwara K, et al.
Scientific Reports, 5 (2015)
18?-Glycyrrhetinic acid interaction with bovine serum albumin.
Zhou N, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 185(2), 271-276 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service