Skip to Content
Merck
All Photos(1)

Documents

H0892

Sigma-Aldrich

Anti-Hemocyanin (KLH) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

keyhole limpet

packaging

antibody small pack of 25 μL

technique(s)

indirect ELISA: 1:20,000

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

General description

Hemocyanin is a large, multi-subunit respiratory protein present in arthropods and mollucs. They circulate extracellularly in hemolymph. Hemocyanins do not contain iron but bind oxygen at an active site containing copper atoms ligated by protein side chains.

Application

Anti-Hemocyanin (KLH) antibody produced in rabbit has been used in:
  • immunocytochemistry{58
  • }immunohistochemistry
  • immunofluorescent staining
  • immunoblotting

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)

Biochem/physiol Actions

Keyhole limpet hemocyanin has been used traditionally as a protein carrier for hapten-carrier conjugates in immunochemistry. Keyhole limpet hemocyanin (KLH) has been regarded as a superior carrier. Affinity isolated antibody to hemocyanin may be used as a control reagent for the preparation and immunization of hapten-carrier conjugates.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dario Armando Leone et al.
Journal of immunology (Baltimore, Md. : 1950), 199(2), 531-546 (2017-06-14)
The lysosome-associated membrane protein (LAMP) family includes the dendritic cell endocytic receptors DC-LAMP and CD68, as well as LAMP-1 and LAMP-2. In this study we identify LAMP-1 (CD107a) and LAMP-2 (CD107b) on the surface of human monocyte-derived dendritic cells (MoDC)
Surface LAMP-2 is an endocytic receptor that diverts antigen internalized by human dendritic cells into highly immunogenic exosomes
Leone DA, et al.
Journal of immunology (Baltimore, Md. : 1950), 199(2), 531-546 (2017)
The murine IL-8 homologues KC, MIP-2, and LIX are found in endothelial cytoplasmic granules but not in Weibel-Palade bodies
Hol J, et al.
Journal of Leukocyte Biology, 87(3), 501-508 (2010)
Nuclear interleukin-33 is generally expressed in resting endothelium but rapidly lost upon angiogenic or proinflammatory activation
Kuchler AM, et al.
The American Journal of Pathology, 173(4), 1229-1242 (2008)
Taylor R Paskin et al.
Behavioural brain research, 235(2), 293-301 (2012-09-12)
The olfactory system is a useful model for studying central nervous system recovery from damage due to its neuroplasticity. We recently developed a novel method of deafferentation by repeated exposure of Triton X-100 to the olfactory organ of adult zebrafish.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service