Skip to Content
Merck
All Photos(1)

Documents

GENA934-1ML

Rabbit IgG HRP Linked Whole Ab

Cytiva NA934-1ML

Synonym(s):

Rabbit IgG Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.76

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

feature

wetted part: no

packaging

pkg of 1 mL

manufacturer/tradename

Cytiva NA934-1ML

color

Brown (light)

shipped in

wet ice

storage temp.

2-8°C

General description

Horseradish peroxidase conjugated antibody is supplied in phosphate buffered saline (sodium phosphate 0.1 M, NaCl 0.1 M) pH 7.5, containing 1% (w/v) bovine serum albumin and an anti-microbial agent.


The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2° C to 8° C.

Features and Benefits

  • Highly species-specific.
  • Optimized for use with our range of ECL Detection Reagents.
  • Recommended dilutions to minimize background.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL is a trademark of Cytiva

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Kelly Dawe et al.
Cell, 173(4), 839-850 (2018-04-10)
Maize abnormal chromosome 10 (Ab10) encodes a classic example of true meiotic drive that converts heterochromatic regions called knobs into motile neocentromeres that are preferentially transmitted to egg cells. Here, we identify a cluster of eight genes on Ab10, called
Shiqi Xiang et al.
Stem cell research & therapy, 12(1), 347-347 (2021-06-16)
Human bone marrow-derived mesenchymal stem cells (hBMSCs) can differentiate into adipocytes upon stimulation and are considered an appropriate cell source for adipose tissue engineering. In addition to biochemical cues, the stiffness of a substrate that cells attach to has also
Baptiste Coxam et al.
Development (Cambridge, England), 149(6) (2022-03-22)
Molecular mechanisms controlling the formation, stabilisation and maintenance of blood vessel connections remain poorly defined. Here, we identify blood flow and the large extracellular protein Svep1 as co-modulators of vessel anastomosis during developmental angiogenesis in zebrafish embryos. Both loss of
Joey De Backer et al.
Scientific reports, 11(1), 10879-10879 (2021-05-27)
Cytoglobin (CYGB) is a ubiquitously expressed protein with a protective role against oxidative stress, fibrosis and tumor growth, shown to be transcriptionally regulated under hypoxic conditions. Hypoxia-inducible CYGB expression is observed in several cancer cell lines and particularly in various
Weixin Xie et al.
Journal of cellular and molecular medicine, 23(5), 3293-3301 (2019-02-19)
Recently, aberrant expression of miR-876-5p has been reported to participate in the progression of several human cancers. However, the expression and function of miR-876-5p in osteosarcoma (OS) are still unknown. Here, we found that the expression of miR-876-5p was significantly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service