Skip to Content
Merck
All Photos(1)

Key Documents

GENA9340-1ML

Rabbit IgG HRP Linked F(ab′)2

Cytiva NA9340-1ML

Synonym(s):

IgG HRP Conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.76
Pricing and availability is not currently available.

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

feature

wetted part: no

packaging

pkg of 1 mL

manufacturer/tradename

Cytiva NA9340-1ML

color

Colorless

shipped in

wet ice

storage temp.

2-8°C

General description

The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2° C to 8° C.

Features and Benefits

  • Highly species-specific.
  • Optimized for use with our range of ECL Detection Reagents.
  • Recommended dilutions to minimize background.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL is a trademark of Cytiva

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service