Skip to Content
Merck
All Photos(2)

Key Documents

G8134

Sigma-Aldrich

Gallamine triethiodide

≥98% (TLC), powder, muscarinic receptor antagonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H60I3N3O3
CAS Number:
Molecular Weight:
891.53
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

mp

235 °C (dec.) (lit.)

solubility

H2O: 100 mg/mL
ethanol: soluble

storage temp.

2-8°C

SMILES string

[I-].[I-].[I-].CC[N+](CC)(CC)CCOc1cccc(OCC[N+](CC)(CC)CC)c1OCC[N+](CC)(CC)CC

InChI

1S/C30H60N3O3.3HI/c1-10-31(11-2,12-3)22-25-34-28-20-19-21-29(35-26-23-32(13-4,14-5)15-6)30(28)36-27-24-33(16-7,17-8)18-9;;;/h19-21H,10-18,22-27H2,1-9H3;3*1H/q+3;;;/p-3

InChI key

REEUVFCVXKWOFE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Gallamine triethiodide has been used:
  • as a relaxant for measuring spinal trigeminal nucleus recordings from single neurons.
  • as an antagonist in neuroblastoma cells as M2 receptor
  • to reduce eye movement during retinal surgery in rat

Biochem/physiol Actions

Gallamine triethiodide has anti-muscarinic effect. It is a competitive antagonist for the muscarinic receptor. Gallamine is regarded as neuromuscular blocking agent.
Muscle relaxant; allosteric muscarinic receptor antagonist with an order of potency of M2 > M1 = M4 > M3 = M5. Ligand for the peripheral anionic binding site of acetylcholinesterase.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Eichler et al.
Molecular pharmacology, 45(2), 335-340 (1994-02-01)
Comparison of the effect of three 'peripheral' site ligands, propidium, d-tubocurarine, and gallamine, on acetylcholinesterase (acetylcholine hydrolase; EC 3.1.1.7) of Torpedo and chicken shows that all three are substantially more effective inhibitors of the Torpedo enzyme than of the chicken
Dar'ya S Redka et al.
Molecular pharmacology, 74(3), 834-843 (2008-06-17)
The M(2) muscarinic receptor has two topographically distinct sites: the orthosteric site and an allosteric site recognized by compounds such as gallamine. It also can exhibit cooperative effects in the binding of orthosteric ligands, presumably to the orthosteric sites within
Neuromuscular blocking drugs: discovery and development
Raghavendra T
Journal of the Royal Society of Medicine, 95(7), 363-367 (2002)
Transcriptional response to muscarinic acetylcholine receptor stimulation: regulation of Egr-1 biosynthesis by ERK, Elk-1, MKP-1, and calcineurin in carbachol-stimulated human neuroblastoma cells
Rossler O, et al.
Archives of Biochemistry and Biophysics, 470(1), 93-102 (2008)
Sandrine B Daval et al.
Molecular pharmacology, 84(1), 71-85 (2013-04-23)
Bitopic binding properties apply to a variety of muscarinic compounds that span and simultaneously bind to both the orthosteric and allosteric receptor sites. We provide evidence that fluorescent pirenzepine derivatives, with the M1 antagonist fused to the boron-dipyrromethene [Bodipy (558/568)]

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service