Skip to Content
Merck
All Photos(1)

Key Documents

G7294

Sigma-Aldrich

Guanidine hydrochloride solution

8 M, pH 8.5, buffered aqueous solution

Synonym(s):

Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, GuHCl, Guanidine HCl, Guanidinium chloride, Guanidium chloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.25

form

buffered aqueous solution

concentration

8 M

pH

8.5

application(s)

diagnostic assay manufacturing

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urea and guanidine hydrochloride denaturation of ribonuclease, lysozyme, alpha-chymotrypsin, and beta-lactoglobulin.
R F Greene et al.
The Journal of biological chemistry, 249(17), 5388-5393 (1974-09-10)
The preparation of guanidine hydrochloride.
Nozaki, Yasuhiko
Methods in Enzymology, 26, 43-50 (1972)
Extended conformations of polypeptides and proteins in urea and guanidine hydrochloride.
Tiffany, M. Lois, and Samuel Krimm
Biopolymers, 12, 575-587 (1973)
Donna Wesolowski et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(40), 16582-16587 (2011-09-29)
Basic peptides covalently linked to nucleic acids, or chemically modified nucleic acids, enable the insertion of such a conjugate into bacteria grown in liquid medium and mammalian cells in tissue culture. A unique peptide, derived from human T cells, has
Davide Gambarotto et al.
Nature methods, 16(1), 71-74 (2018-12-19)
Determining the structure and composition of macromolecular assemblies is a major challenge in biology. Here we describe ultrastructure expansion microscopy (U-ExM), an extension of expansion microscopy that allows the visualization of preserved ultrastructures by optical microscopy. This method allows for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service