Skip to Content
Merck
All Photos(4)

Documents

G5509

Sigma-Aldrich

D-Glucosamine 6-phosphate

≥98% (TLC)

Synonym(s):

2-Amino-2-deoxyglucose 6-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14NO8P
CAS Number:
Molecular Weight:
259.15
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

natural (inorganic)

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

impurities

≤10% water (Karl Fischer)

color

white

solubility

water: 50 mg/mL, clear to hazy, colorless to almost colorless

storage temp.

−20°C

SMILES string

NC(C=O)C(O)C(O)C(O)COP(O)(O)=O

InChI

1S/C6H14NO8P/c7-3(1-8)5(10)6(11)4(9)2-15-16(12,13)14/h1,3-6,9-11H,2,7H2,(H2,12,13,14)

InChI key

AEJSSXDYDSUOOZ-UHFFFAOYSA-N

General description

D-Glucosamine 6-phosphate, the natural form of glucosamine, is a monosaccharide produced during hexosamine biosynthesis pathway by the enzyme glucosamine-6-phosphate deaminase from the two substrates fructose 6-phosphate and glutamine.

Linkage

Stability is greater than that of the sodium salt.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanislav S Terekhov et al.
Science advances, 6(26), eaaz9861-eaaz9861 (2020-07-09)
Microbial communities are self-controlled by repertoires of lethal agents, the antibiotics. In their turn, these antibiotics are regulated by bioscavengers that are selected in the course of evolution. Kinase-mediated phosphorylation represents one of the general strategies for the emergence of
Reflections on glycobiology.
S Roseman
The Journal of biological chemistry, 276(45), 41527-41542 (2001-09-13)
James S Davies et al.
FEBS letters, 593(1), 52-66 (2018-11-10)
N-Acetylglucosamine-6-phosphate deacetylase (NagA) and glucosamine-6-phosphate deaminase (NagB) are branch point enzymes that direct amino sugars into different pathways. For Staphylococcus aureus NagA, analytical ultracentrifugation and small-angle X-ray scattering data demonstrate that it is an asymmetric dimer in solution. Initial rate experiments
Mark E Graham et al.
Organic & biomolecular chemistry, 10(13), 2545-2551 (2012-03-01)
A novel post-translational modification of threonine, β-N-acetylglucosaminyl-phosphate, was recently discovered on assembly protein AP180, a protein which plays a crucial role in clathrin coated vesicle formation in synaptic vesicle endocytosis (SVE). Herein, we report studies aimed at probing the effect
Marie Valerio-Lepiniec et al.
Archives of biochemistry and biophysics, 498(2), 95-104 (2010-04-27)
Glucosamine-6-phosphate synthase (GlmS) is responsible for the first and rate-limiting step in the hexosamine biosynthetic pathway. It catalyzes the conversion of D-fructose-6P (F6P) into D-glucosamine-6P (GlcN6P) using L-glutamine (Gln) as nitrogen donor (synthase activity) according to an ordered bi-bi process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service