Skip to Content
Merck
All Photos(2)

Key Documents

286583

Sigma-Aldrich

5-Methoxytryptamine

97%

Synonym(s):

2-(5-Methoxyindol-3-yl)ethylamine, 3-(2-Aminoethyl)-5-methoxyindole, 5-MOT, 5-Methoxyindole-3-ethanamine, Deacetylmelatonin, NSC 56422

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
Beilstein:
145587
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

121-123 °C (lit.)

functional group

amine

SMILES string

COc1ccc2[nH]cc(CCN)c2c1

InChI

1S/C11H14N2O/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11/h2-3,6-7,13H,4-5,12H2,1H3

InChI key

JTEJPPKMYBDEMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The protective effect of 5-methoxytryptamine (a metabolite of melatonin) in human keratinocytes against ultraviolet B (UVB) radiation was studied.

Application

5-Methoxytryptamine was used as an agonist in the study of pharmacological profile of the 5-hydroxytryptamine 1 receptor.
Reactant for preparation of:
  • Carboline disaccharide domain of shishijimicin A
  • Melatonin analogs for the reduction of intraocular pressure
  • 5-HT4 receptor ligands
  • inhibitors of sortase A and isocitrate lyase
  • Therapeutic agents for treatment of ischemia/reperfusion (I/R) injury
  • Aurora and epidermal growth factor receptor kinase inhibitors
  • Agents for the treatment of human papillomavirus infection
  • Manzamine analogues for the control of neuroinflammation and cerebral infections
  • Inhibitors of pro-inflammatory cytokines
  • Tacrine-melatonin hybrids as multifunctional agents for alzheimer′s disease

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Amir Hanna-Elias et al.
European journal of medicinal chemistry, 44(7), 2952-2959 (2009-02-19)
Twenty-three indole-3-methanamines were designed, synthesized and evaluated as ligands for the 5-HT(4) receptor. Compounds I-d, I-j, I-o, I-q and I-u showed good affinity at 100 microM and I-o was found to be only 5-fold less potent than the agonists serotonin
Melatonin and its metabolites ameliorate ultraviolet B-induced damage in human epidermal keratinocytes.
Janjetovic Z, Nahmias ZP, Hanna S, et al.
Journal of Pineal Research, 57(1), 90-102 (2014)
Hyun-Ji Kim et al.
Combinatorial chemistry & high throughput screening, 11(4), 316-324 (2008-05-14)
Continuous identification and validation of novel drug targets require the development of rapid, reliable, and sensitive cell-based high-throughput screening (HTS) methods for proposed targets. Recently, the 5-HT(6) receptor (5-HT(6)R), a member of the class of recently discovered 5-HT receptors, has
Anna Haduch et al.
Pharmacological research, 67(1), 53-59 (2012-10-27)
Brain serotonin has been implicated in the pathophysiology of a wide spectrum of psychiatric disorders, as well as in the mechanism of action of psychotropic drugs. The aim of present study was to identify rat cytochrome P450 (CYP) isoforms which
Jeff DeFalco et al.
Bioorganic & medicinal chemistry letters, 20(23), 7076-7079 (2010-10-23)
5-Benzyloxytryptamine 19 was found to act as an antagonist of the TRPM8 ion-channel. For example, 19 had an IC(50) of 0.34 μM when menthol was used as the stimulating agonist. Related commercially-available tryptamine derivatives showed diminished, or no antagonist activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service