Skip to Content
Merck
All Photos(5)

Documents

G1N10

Sigma-Aldrich

GenElute Mammalian Genomic DNA Miniprep Kits

sufficient for 10 purifications

Synonym(s):

Mammalian Genomic DNA Miniprep, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

usage

sufficient for 10 purifications

Quality Level

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Mammalian Genomic DNA Purification Kit provides a simple and convenient way to isolate pure, high molecular weight DNA from a variety of mammalian sources. These kits use a silica-based membrane, specially selected for genomic DNA purification, in a convenient spin column format.

Isolate pure genomic DNA from cultured cells, tissues (including rodent tails), and fresh whole blood or white blood cells. The kit combines the advantages of silica binding with a microspin format and eliminates the need for expensive resins, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform.

Application

The purified genomic DNA is ready for immediate use in downstream applications such as:
  • restriction endonuclease digestions
  • PCR
  • Southern blots
  • sequencing reactions
  • cloning
  • ligation

Features and Benefits

  • Expected yield: 25 μg from 2 x 106 cultured cells; 30 μg from 25 mg of tissue
  • Elution volume: 200 - 400 μl
  • Time required: 20 min after lysis
  • A260/A280 ratio: 1.6 - 1.9
  • Mechanical homogenization required: No

Principle

The starting materials are lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. Addition of ethanol causes DNA to bind when the lysate is spun through a silica membrane in a microcentrifuge tube. After washing to remove the contaminants, the DNA is eluted in 200 μL of a Tris-EDTA solution. The expected yields of genomic DNA will vary depending on the amount and type of starting material used. DNA purified with the kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • C2112Column Preparation SolutionSDS

  • P2308Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biologySDS

  • R6148RNase A solutionSDS

recommended

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

483.8 °F

Flash Point(C)

251 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Züleyha Doğanyiğit et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 20(7), 632-639 (2013-03-05)
In recent years, propolis has been the object of extensive research for its antibacterial, antioxidant, anti-inflammatory, and antitumoral activities. This study aims to determine the hepatoprotective efficiency of propolis on experimental endotoxemia in rats. In the current study, fifty adult
Michal Kirshner et al.
Journal of molecular neuroscience : MN, 46(3), 554-568 (2011-09-17)
Pronounced neuropathology is a feature of ataxia-telangiectasia (A-T) and Nijmegen breakage syndrome (NBS), which are both genomic instability syndromes. The Nbs1 protein, which is defective in NBS, is a component of the Mre11/RAD50/NBS1 (MRN) complex. This complex plays a major
Giulia Crispino et al.
Scientific reports, 7(1), 6567-6567 (2017-08-06)
We have previously shown that in vitro transduction with bovine adeno-associated viral (BAAV) vectors restores connexin expression and rescues gap junction coupling in cochlear organotypic cultures from connexin-deficient mice that are models DFNB1 nonsyndromic hearing loss and deafness. The aims
Natalia N Pouchkina-Stantcheva et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 138(4), 371-376 (2004-08-25)
Various spider species produce dragline silks with different mechanical properties. The primary structure of silk proteins is thought to contribute to the elasticity and strength of the fibres. Previously published work has demonstrated that the dragline silk of Euprosthenops sp.
John B A Okello et al.
Analytical biochemistry, 400(1), 110-117 (2010-01-19)
Archival formalin-fixed paraffin-embedded (FFPE) human tissue collections are typically in poor states of storage across the developing world. With advances in biomolecular techniques, these extraordinary and virtually untapped resources have become an essential part of retrospective epidemiological studies. To successfully

Protocols

Archived Formalin-fixed, Paraffin-embedded (FFPE) tissue samples are invaluable resources for profiling gene expression and studying a variety of diseases.

Archived Formalin-fixed, Paraffin-embedded (FFPE) tissue samples are invaluable resources for profiling gene expression and studying a variety of diseases.

Archived Formalin-fixed, Paraffin-embedded (FFPE) tissue samples are invaluable resources for profiling gene expression and studying a variety of diseases.

Archived Formalin-fixed, Paraffin-embedded (FFPE) tissue samples are invaluable resources for profiling gene expression and studying a variety of diseases.

See All

Related Content

GenElute Mammalian Genomic DNA Purification Kit isolates pure DNA from mammalian cells using a silica-based membrane in spin columns.

GenElute Mammalian Genomic DNA Purification Kit isolates pure DNA from mammalian cells using a silica-based membrane in spin columns.

GenElute Mammalian Genomic DNA Purification Kit isolates pure DNA from mammalian cells using a silica-based membrane in spin columns.

GenElute Mammalian Genomic DNA Purification Kit isolates pure DNA from mammalian cells using a silica-based membrane in spin columns.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service