Skip to Content
Merck
All Photos(1)

Documents

G1146

Sigma-Aldrich

ʟ-Glutamine–Penicillin–Streptomycin

solution, suitable for cell culture, BioReagent

Synonym(s):

Pen/Strep, Penicillin/Streptomycin 

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.76

product name

L-Glutamine–Penicillin–Streptomycin solution, with 200 mM L-glutamine, 10,000 U penicillin and 10 mg steptomycin/mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture

biological source

synthetic

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes
protein synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Used to reduce or eliminate bacterial contamination in cell, especially primary cell, cultures. Recommended for use in cell culture applications at 10 ml/L.

Biochem/physiol Actions

Mode of Action: Glutamine is essential as an energy source for most mammalian cells in culture because of its amino acid carbon skeleton. Penicillin acts by inhibiting bacterial cell wall synthesis. Streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome and causes miscoding.
Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.

Components

Formulated with 200 mM L-glutamine, 10,000 units/ml penicillin, and 10 mg/ml streptomycin in 0.9% sodium chloride.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Vcelakova et al.
Journal of diabetes research, 2013, 589451-589451 (2013-07-11)
Type 1 Diabetes (T1D) is considered to be a T-helper- (Th-) 1 autoimmune disease; however, T1D pathogenesis likely involves many factors, and sufficient tools for autoreactive T cell detection for the study of this disease are currently lacking. In this
Fumito Tamura et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 19(1), 85-97 (2014-12-24)
ST6GalNAc I is a sialyltransferase controlling the expression of sialyl-Tn antigen (STn), which is overexpressed in several epithelial cancers, including gastric cancer, and is highly correlated with cancer metastasis. However, the functional contribution of ST6GalNAc I to development or progression
Chi-Fei Wang et al.
Anesthesia and analgesia, 107(4), 1397-1405 (2008-09-23)
Bulleyaconitine A (BLA) is an analgesic and antiinflammatory drug isolated from Aconitum plants. BLA has several potential targets, including voltage-gated Na+ channels. We tested whether BLA elicited long-lasting cutaneous analgesia, when co-injected with lidocaine and epinephrine, as a model for
Ulrich Wernery et al.
PloS one, 5(12), e15824-e15824 (2011-01-07)
The Houbara bustard (Chlamydotis undulata) is a wild seasonal breeding bird populating arid sandy semi-desert habitats in North Africa and the Middle East. Its population has declined drastically during the last two decades and it is classified as vulnerable. Captive
Angeliki Ditsiou et al.
Science advances, 6(46) (2020-11-15)
Elucidating signaling driven by lemur tyrosine kinase 3 (LMTK3) could help drug development. Here, we solve the crystal structure of LMTK3 kinase domain to 2.1Å resolution, determine its consensus motif and phosphoproteome, unveiling in vitro and in vivo LMTK3 substrates.

Articles

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service