Skip to Content
Merck
All Photos(1)

Key Documents

F0805

Sigma-Aldrich

Anti-FOXL2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Forkhead L2, Anti-Forkhead Transcription Factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, hamster

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 0.2 μg/mL using murine prenatal folicle
indirect immunofluorescence: 2 μg/mL using KK1 granulosa cells
western blot: 0.2 μg/mL using CHO whole cell lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Foxl2(26927)

Immunogen

synthetic peptide M(1)MASYPEPEDTAGT(14) corresponding to amino acid residues 1-14 from mouse FOXL2. This sequence is completely conserved in mouse and rat and 92% conserved in human.

Application

Anti-FOXL2 antibody produced in rabbit is suitable for the following applications:
  • Immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 0.2μg/mL using murine prenatal folicle
  • Indirect immunofluorescence at a concentration of 2μg/mL using KK1 granulosa cells
  • Western blotting (at a concentration of 0.2μg/mL using CHO whole cell lysate) or immunoblot analysis.

Biochem/physiol Actions

Forkhead box protein L2 is a protein encoded by the FOXL2 gene in humans. The gene FOXL2 encodes a member of the fork-head-winged-helix family of transcription factors. It is expressed early during female gonadal development and is expressed in normal ovarian stroma and ovarian neoplasms with granulosa cell lineage. FOXL gene encodes an essential transcription factor in the ovary and helps in female sex determination, follicle recruitment and granulosa cell development. Its mutation is specific to AGCTs in the ovary and is useful for diagnosis of disease. FOXL2 acts as an oncogene or tumour suppressor depending on the genetic context that is the GCT subtype.

Target description

FOXL2 encodes a forkhead transcription factor. The protein contains a fork-head DNA-binding domain and may play a role in ovarian development and function. Mutations in this gene are a cause of blepharophimosis syndrome and premature ovarian failure 3.

Physical form

Provided as 100 μg of affinity purified IgG (1 mg/mL) in phosphate buffered saline containing 1 mg/mL bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Westerhoff et al.
Human pathology, 45(5), 1010-1014 (2014-04-22)
FOXL2, a gene encoding a member of the fork-head-winged-helix family of transcription factors, is one of the earliest expressed genes during female gonadal development. It is expressed in normal ovarian stroma and ovarian neoplasms with granulosa cell lineage. Nonovarian tumors
Fubiao Shi et al.
Human molecular genetics, 23(14), 3792-3800 (2014-02-26)
Blepharophimosis, ptosis, epicanthus inversus syndrome (BPES) is an autosomal dominant genetic disorder characterized by small palpebral fissures and other craniofacial malformations, often with (type I) but could also without (type II) premature ovarian failure. While mutations of the forkhead transcription
Roseanne Rosario et al.
Gynecologic oncology, 133(2), 382-387 (2013-12-18)
It has been four years since the discovery of the FOXL2 402C>G mutation in adult ovarian granulosa cell tumours. Yet to date, there have been few studies which have investigated the precise role of the mutation in tumour pathogenesis. This
Akimasa Takahashi et al.
The Tohoku journal of experimental medicine, 231(4), 243-250 (2013-11-22)
Adult-type granulosa cell tumor (AGCT) is a rare class of malignant ovarian tumor with unique features, characterized by slow growth, late recurrence, relatively good prognosis and unified cause in almost all patients. The forkhead box L2 (FOXL2) gene encodes an
Adrien Levasseur et al.
Biology of reproduction, 97(1), 162-175 (2017-06-24)
Yes-associated protein (YAP) and WW-containing transcription regulator 1 (WWTR1) are two functionally redundant transcriptional regulators that are downstream effectors of the Hippo signaling pathway, and that act as major regulators of cell growth and differentiation. To elucidate their role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service