EP0030108450
Eppendorf® Protein LoBind Tubes
PCR clean, withstand 18,000 x g, 2.0 mL, pack of 500 ea
Synonym(s):
Eppendorf® Protein LoBind tubes
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Recommended Products
material
colorless
conical bottom
description
Low retention
sterility
non-sterile
feature
DNase free
PCR clean
packaging
pack of 500 ea
manufacturer/tradename
Eppendorf® 0030108450
capacity
2.0 mL
suitability
suitable for PCR
Related Categories
General description
Avoid losing valuable samples during centrifugation, incubation and storage! Rely on the excellent protection offered by the hinged Safe-Lock lid. It contains all of our experience from 50 years of continuous optimization and development. Trust the original Eppendorf Safe-Lock Tubes, because your samples deserve only the best.
Features and Benefits
Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)