EP0030122240
Eppendorf® Protein LoBind Tubes
capacity 50 mL, PCR clean, pack of 200 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
Recommended Products
material
colorless tube
conical bottom
screw top cap
white cap
sterility
non-sterile
feature
PCR clean
graduations
neck thread External thread
packaging
pack of 200 ea
manufacturer/tradename
Eppendorf® 0030122240
technique(s)
PCR: suitable
capacity
50 mL
binding type
low binding surface
Looking for similar products? Visit Product Comparison Guide
Application
- Preparation or storage of protein, peptide or antibody samples.
- Enzymatic assays – the hydrophilic surface reduces denaturation when it comes into contact with the tube inner wall.
- Prevention of sample loss during storage of virus stock solutions.
- Storage of cell suspensions.
Features and Benefits
- Eppendorf® LoBind material ensures optimized sample recovery for improved assay results.
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- Available in tube formats for easy-up scaling.
- Precise lid sealing to minimize evaporation.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service