Skip to Content
Merck
All Photos(1)

Key Documents

EHU107721

Sigma-Aldrich

MISSION® esiRNA

targeting human HSF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATTCCCTCCTTGCTCGAGATGGATCTGCCCGTGGGCCCCGGCGCGGCGGGGCCCAGCAACGTCCCGGCCTTCCTGACCAAGCTGTGGACCCTCGTGAGCGACCCGGACACCGACGCGCTCATCTGCTGGAGCCCGAGCGGGAACAGCTTCCACGTGTTCGACCAGGGCCAGTTTGCCAAGGAGGTGCTGCCCAAGTACTTCAAGCACAACAACATGGCCAGCTTCGTGCGGCAGCTCAACATGTATGGCTTCCGGAAAGTGGTCCACATCGAGCAGGGCGGCCTGGTCAAGCCAGAGAGAGACGACACGGAGTTCCAGCACCCATGCTTCCTGCGTGGCCAGGAGCAGCTCCTTGAGAACATCAAGAGGAAAGTGACCAGTGTGTCCACCCTGAAGAGTGAAGACATAAAGATCCGCCAGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Cigliano et al.
Oncotarget, 8(33), 54149-54159 (2017-09-15)
Upregulation of the heat shock transcription factor 1 (HSF1) has been described as a frequent event in many cancer types, but its oncogenic role in hepatocellular carcinoma (HCC) remains poorly delineated. In the present study, we assessed the function(s) of
Liqun Shang et al.
Life sciences, 241, 117120-117120 (2019-12-12)
The present study explored the function and regulatory mechanism of High mobility group box 1 (HMGB1) in asthma. OVA (ovalbumin)-induced asthmatic mice model and LPS-treated cellular model were established in this study. Airway inflammation was measured through detecting the expression
Seok Jun Kim et al.
Yonsei medical journal, 59(9), 1041-1048 (2018-10-18)
Heat shock factor 1 (HSF1) is a key regulator of the heat shock response and plays an important role in various cancers. However, the role of HSF1 in gastric cancer is still unknown. The present study evaluated the function of
Liang Chen et al.
Cell cycle (Georgetown, Tex.), 18(1), 60-68 (2018-12-20)
Cells mainly rely on stress proteins, such as heat-shock proteins (HSPs), to respond to various proteotoxic conditions. These proteins protect tumor cells and enhance their survive. However, the regulation of stress proteins involved in protein quality control (PQC) is still
Ye-Ji Jeong et al.
PloS one, 10(6), e0128552-e0128552 (2015-06-02)
Radiation enteropathy is a common complication in cancer patients. The aim of this study was to investigate whether radiation-induced intestinal injury could be alleviated by coniferyl aldehyde (CA), an HSF1-inducing agent that increases cellular HSP70 expression. We systemically administered CA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service