Skip to Content
Merck
All Photos(1)

Key Documents

EHU000311

Sigma-Aldrich

MISSION® esiRNA

targeting human IRF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGTGTACCGGATGCTTCCACCTCTCACCAAGAACCAGAGAAAAGAAAGAAAGTCGAAGTCCAGCCGAGATGCTAAGAGCAAGGCCAAGAGGAAGTCATGTGGGGATTCCAGCCCTGATACCTTCTCTGATGGACTCAGCAGCTCCACTCTGCCTGATGACCACAGCAGCTACACAGTTCCAGGCTACATGCAGGACTTGGAGGTGGAGCAGGCCCTGACTCCAGCACTGTCGCCATGTGCTGTCAGCAGCACTCTCCCCGACTGGCACATCCCAGTGGAAGTTGTGCCGGACAGCACCAGTGATCTGTACAACTTCCAGGTGTCACCCATGCCCTCCACCTCTGAAGCTACAACAGATGAGGATGAGGAAGGGAAATTACCTGAGGACATCATGAAGCTCTTGGAGCAGTCGGAGTGGCAGCCAACAAACGTGGATGGGAAGGGGTACCTACTCAATGAACCTGGAGTCCAGCCCACCTCTGTCTATGGAGACTTTAGCTGTAAGGAGGAGCCAGAAATTGACAGCCCAGGGGGGGATATTGGGCTGAGTCTACAGCGTGTCTTCACAGATCTGAAGAACATGGATGCCACCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yihe Yan et al.
Cancer immunology, immunotherapy : CII, 69(9), 1891-1903 (2020-05-08)
The objective response rate of immune checkpoint blockade (ICB) in hepatocellular carcinoma (HCC) with anti PD-L1/PD-1 therapy is low. Discovering the signaling pathways regulating PD-L1 might help to improve ICB response rates. Here, we investigate transcription factors IRF-1 and IRF-2
Z-D Liu et al.
European review for medical and pharmacological sciences, 24(23), 12334-12341 (2020-12-19)
Cerebral ischemia/reperfusion (CIR) frequently causes serious disabilities and correlates with certain neurological processes. Some studies have shown that microRNAs (miRNAs) exert a neuroprotective effect by modulating the inflammatory process in CIR. However, the biofunction and the mechanism of miR-130b in
Eri Sugiyama et al.
Science immunology, 5(43) (2020-02-02)
The clinical efficacy of anti-PD-1 (programmed cell death-1) monoclonal antibody (mAb) against cancers with oncogenic driver gene mutations, which often harbor a low tumor mutation burden, is variable, suggesting different contributions of each driver mutation to immune responses. Here, we
Lulu Tan et al.
American journal of cancer research, 10(4), 1255-1270 (2020-05-06)
Recent studies have shown that IRF-1 plays a significant role in various tumour-induced chemoresistance, but its role and mechanism in gastric cancer-associated chemoresistance are not clear. Our study showed that IRF-1 expression could reverse gastric cancer-related chemoresistance. Dysregulated DNA repair
Meng Du et al.
Theranostics, 9(16), 4688-4703 (2019-08-02)
Deciphering the molecular and cellular processes involved in foam cell formation is critical to understanding the pathogenesis of atherosclerosis. Interferon regulatory factor 1 (IRF1) was first identified as a transcriptional regulator of type-I interferons (IFNs) and IFN inducible genes. Our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service