Skip to Content
Merck
All Photos(4)

Key Documents

EDFS

Sigma-Aldrich

Ethylenediaminetetraacetic acid iron(III) sodium salt

powder

Synonym(s):

EDTA iron(III) sodium salt, Edathamil, Komplexon II

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12N2NaFeO8
CAS Number:
Molecular Weight:
367.05
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Agency

suitable for SM 5210

form

powder

composition

Iron, 13.7-18.7% ICP
Sodium, 5.6-7.9% ICP

reaction suitability

reagent type: chelator

concentration

≥67.5% (EDTA titration)

pH

4.5

mp

80 °C

solubility

water: 50 mg/mL, clear to slightly hazy, yellow to brown

storage temp.

room temp

SMILES string

[Na+].[Fe+3].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Fe.Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;+3;+1/p-4

InChI key

MKWYFZFMAMBPQK-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. Iron-EDTA enhances damage due to reactive oxygen species, such as depletion of reduced glutathione, formation of methemoglobin, and lipid peroxidation.

Legal Information

Komplexon is a trademark of CU Chemie Uetikon AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasha T Forester et al.
Molecular plant-microbe interactions : MPMI, 32(10), 1324-1335 (2019-05-21)
In ascomycetes and basidiomycetes, iron-responsive GATA-type transcriptional repressors are involved in regulating iron homeostasis, notably to prevent iron toxicity through control of iron uptake. To date, it has been unknown whether this iron regulator contributes toward mutualistic endosymbiosis of microbes
María J Binaghi et al.
Archivos latinoamericanos de nutricion, 61(3), 316-322 (2012-06-16)
Home-made diets are the most frequently used complementary foods. In the present work we evaluated iron and zinc availability in a usually consumed infant diet containing either iron-fortified bread with different iron sources: ferrous sulfate, ferrous bisglycinate, NaFeEDTA. We also
Bo Wang et al.
Wei sheng yan jiu = Journal of hygiene research, 40(3), 334-337 (2011-06-24)
To assess the effectiveness of social mobilization and social marketing in promoting NaFeEDTA-fortified soy sauce in an iron deficient population. This study was an uncontrolled, community-based, before-after study, which was implemented in three counties of Shijiazhuang Municipality. The intervention was
Suying Chang et al.
Food and nutrition bulletin, 33(2), 111-116 (2012-08-23)
Ferric sodium ethylenediaminetetraacetate (NaFeEDTA) enhances iron absorption in the presence of phytate. However, the amount of NaFeEDTA that would have to be added to a complementary food to provide the necessary intake of iron for an infant or young child
Monica N Orozco et al.
Annals of nutrition & metabolism, 60(2), 108-114 (2012-03-15)
In any context of iron supplementation in the prenatal prophylaxis or therapeutic dosage range, a large amount will remain unabsorbed and pass through the intestinal tract into the colonic digesta possibly causing increased oxidation. To compare the generation of fecal

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service