Skip to Content
Merck
All Photos(1)

Key Documents

E9012

Sigma-Aldrich

Endothelin 2 human

≥97% (HPLC), powder

Synonym(s):

ET-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C115H160N26O32S4
CAS Number:
Molecular Weight:
2546.92
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@@H]2CSSC[C@H](N)C(=O)N[C@@H](CO)C(=O)N[C@H]3CSSC[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc4c[nH]c5ccccc45)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC3=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc6ccc(O)cc6)C(=O)N[C@@H](Cc7ccccc7)C(=O)N2)[C@@H](C)CC)C(=O)N[C@@H](Cc8c[nH]c9ccccc89)C(O)=O

InChI

1S/C115H160N26O32S4/c1-11-59(9)93(113(170)132-82(115(172)173)41-64-46-120-71-27-19-17-25-68(64)71)141-114(171)94(60(10)12-2)140-105(162)81(44-91(150)151)130-99(156)75(37-57(5)6)125-103(160)79(42-65-47-118-55-121-65)128-109(166)86-52-175-174-51-69(117)95(152)133-83(48-142)108(165)138-87-53-176-177-54-88(111(168)139-92(58(7)8)112(169)131-77(39-62-29-31-66(145)32-30-62)100(157)126-76(101(158)137-86)38-61-22-14-13-15-23-61)136-97(154)73(33-34-89(146)147)123-96(153)72(28-20-21-35-116)122-104(161)80(43-90(148)149)129-98(155)74(36-56(3)4)124-102(159)78(40-63-45-119-70-26-18-16-24-67(63)70)127-106(163)84(49-143)134-107(164)85(50-144)135-110(87)167/h13-19,22-27,29-32,45-47,55-60,69,72-88,92-94,119-120,142-145H,11-12,20-21,28,33-44,48-54,116-117H2,1-10H3,(H,118,121)(H,122,161)(H,123,153)(H,124,159)(H,125,160)(H,126,157)(H,127,163)(H,128,166)(H,129,155)(H,130,156)(H,131,169)(H,132,170)(H,133,152)(H,134,164)(H,135,167)(H,136,154)(H,137,158)(H,138,165)(H,139,168)(H,140,162)(H,141,171)(H,146,147)(H,148,149)(H,150,151)(H,172,173)/t59-,60-,69-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,92-,93-,94-/m0/s1

InChI key

MLFJHYIHIKEBTQ-IYRKOGFYSA-N

Gene Information

human ... EDN2(1907)

General description

Endothelin-2 (ET-2) is a member of the endothelin family, which also includes ET-1 and ET-3. ET-2 is the human ortholog of mouse and rat vasoactive intestinal contractor (VIC). These peptides are characterized by two disulfide linkages. ET-2 is predominantly expressed in cerebrum, stomach, intestine, ovary, uterus and testis.

Biochem/physiol Actions

Endothelin-2 (ET-2) has a regulatory role in multiple cancer types, such as breast and clear cell renal cell carcinoma (ccRCC). It is overexpressed during early stages of non-metastasized ccRCC, and is thought have a role in the progression of the same. It is a hypoxia-related factor, and might have protective action against cell death in neuronal diseases. It acts as a ligand for ET-A and ET-B receptors, which results in vasoconstriction. A985 polymorphism in this gene is thought play a protective role in cardiovascular diseases, but it acts as a susceptibility factor in atrial fibrillation in hypertrophic cardiomyopathy patients. In neural retina, it suppresses angiogenesis, and facilitates the tip cell state in endothelial cells.

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian M Bot et al.
Human pathology, 43(6), 843-849 (2011-11-04)
Despite the rising incidence of clear cell renal cell carcinoma, the molecular events that support its development and progression remain unclear. Herein, we evaluate the association of endothelin 2 expression with both clear cell renal cell carcinoma development and progression-free
G L Lambert et al.
Journal of molecular endocrinology, 24(2), 273-283 (2000-04-06)
ABSTRACT The synthesis of the vasoconstrictor peptide endothelin-2 (ET-2) is dependent on hydrolysis of the biologically inactive intermediate big ET-2 by an endothelin-converting enzyme (ECE). Here, mechanisms inducing ET-2 synthesis have been investigated using the human renal adenocarcinoma cell line
Amir Rattner et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(40), E3830-E3839 (2013-09-18)
Endothelin signaling is required for neural crest migration and homeostatic regulation of blood pressure. Here, we report that constitutive overexpression of Endothelin-2 (Edn2) in the mouse retina perturbs vascular development by inhibiting endothelial cell migration across the retinal surface and
Premkumari Kumarathasan et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 21(3), 257-266 (2016-02-24)
There is paucity of information on mechanisms constituting adverse birth outcomes. We assessed here the relationship between vascular integrity and adverse birth effects. Third trimester maternal plasma (n = 144) from the Maternal-Infant Research on Environmental Chemicals Study (MIREC) was
Eiichi Kotake-Nara et al.
TheScientificWorldJournal, 6, 176-186 (2006-02-24)
This paper reviews the local hormone endothelin-2 (ET-2), or vasoactive intestinal contractor (VIC), a member of the vasoconstrictor ET peptide family, where ET-2 is the human orthologous peptide of the murine VIC. While ET-2/VIC gene expression has been observed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service