Skip to Content
Merck
All Photos(3)

Key Documents

E6375

Sigma-Aldrich

Estriol 3-sulfate sodium salt

≥98% (TLC)

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17β-triol 3-sulfate, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene 3-sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H23O6SNa
CAS Number:
Molecular Weight:
390.43
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
form:
powder
Assay:
≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

contains

~30% methylglucamine as stabilizer

solubility

methanol: 19.60-20.40 mg/mL, clear, colorless to faintly yellow

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

room temp

SMILES string

[Na].[H][C@]12CC[C@]3(C)[C@@H](O)[C@H](O)C[C@@]3([H])[C@]1([H])CCc4cc(OS(O)(=O)=O)ccc24

InChI

1S/C18H24O6S.Na.H/c1-18-7-6-13-12-5-3-11(24-25(21,22)23)8-10(12)2-4-14(13)15(18)9-16(19)17(18)20;;/h3,5,8,13-17,19-20H,2,4,6-7,9H2,1H3,(H,21,22,23);;/t13-,14-,15+,16-,17+,18+;;/m1../s1

InChI key

NGMCHEUPKPRHNC-XZEIGEQDSA-N

Packaging

Package size based on steroid content

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

U Raju et al.
Steroids, 50(4-6), 559-574 (1987-10-01)
The concentration of 16 alpha-hydroxydehydroepiandrosterone-3-sulfate (16 alpha-OHDHAS) was determined in 29 samples of human breast cyst fluid (BCF) and in 15 of these, androst-5-ene-3 beta,16 alpha,17 beta-triol-3-sulfate (A-TriolS) was also assayed. The median value of both was about 100 ng/mL
N Tagawa et al.
Biological & pharmaceutical bulletin, 21(11), 1211-1214 (1998-12-16)
A method to prevent co-elution of steroid sulfates with proteins in serum from the pre-column in column-switching HPLC was developed. The pre-column, a polymer-coated mixed function column, was used for ion-pair chromatography with 5 mM tetra-n-butylammonium (TBA) ion. As steroid
U Raju et al.
Steroids, 45(3-4), 341-346 (1985-03-01)
Estriol-3-sulfate (E3-3S) was assayed in 92 specimens of human breast cyst fluid (BCF) obtained by needle aspiration from women with fibrocystic disease. The concentrations of K+ and Na+ were determined in the same samples. The median concentration of E3-3S in
U Raju et al.
Journal of steroid biochemistry, 20(4B), 1061-1065 (1984-04-01)
Although cystic lesions of the breast are not precancerous per se, statistical studies have indicated that this condition predisposes a 2- to 4-fold greater risk for breast cancer. Seeking a hormonal etiology to this correlation, investigators have analyzed breast cyst
U Raju et al.
The Journal of clinical endocrinology and metabolism, 53(4), 847-851 (1981-10-01)
The high concentration of estriol-3-sulfate (E3-3S) in human breast cyst fluid has been confirmed in 14 women with multiple cysts. The concentrations of E3-3S found in individual cysts drained within a short time span from the same patient were variable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service