Skip to Content
Merck
All Photos(4)

Key Documents

D8821

Sigma-Aldrich

Dextran from Leuconostoc mesenteroides

average mol wt 60,000-76,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

bacterial (Leuconostoc mesenteroides)

form

powder

optical activity

[α]20/D 190 to 201 °, c = 2% (w/v) in water

mol wt

average mol wt 60,000-76,000

color

white to off-white

mp

483  °C ((901 °F ))

solubility

water: 100 mg/mL at 25 °C, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dextran is an extracellular bacterial polymer of D-glucopyranose. It consists of α-(1→6) linkage in the main chain with a variable amount of α-(1→2), α-(1→3), α-(1→4) branched linkages. Dextran from L. mesenteroides B512F comprises 95% of α- (1→6) linkages and 5% of α-(1→3) branch linkages. Insoluble dextran from L. mesenteroides 1299 contains 63% α-(1→6), 27% of α-(1→2) and 8% of α-(1→3) linkages.

Application

Dextran from Leuconostoc mesenteroides has been used:
  • in resuspension of pellets and in Hanks′ balanced salt solution (HBSS) for the isolation of vascular nuclei from frozen post-mortem brain tissue
  • to prepare dextran solution in phosphate buffer saline and to maintain corneal hydration and restore corneal thickness to physiological levels of human eyes
  • as a polymer solution in the in situ aqueous two-phase systems (ATPS) formation with lysozyme PEGylation reaction

Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules.

Biochem/physiol Actions

Dextran is commercially available as a drug, especially as a blood plasma volume expander. This bacterial polysaccharide has several applications, such as adjuvant, emulsifier, carrier, and stabilizer in the food, pharmaceutical, and chemical industries. Dextran is used in the thickening of jam and ice cream in the food industry. It prevents sugar crystallization and retains moisture. This helps in preserving the flavor and appearance of several food items.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janice M Dias et al.
Experimental eye research, 115, 41-46 (2013-06-27)
Corneal biomechanics is an essential parameter for developing diagnostic and treatment methods of corneal-related diseases. It is widely accepted that corneal mechanical strength stems from the stroma's collagenous composition. However, more comprehensive insight into the mechanical properties within the stroma
Farwa Sarwat et al.
International journal of biological sciences, 4(6), 379-386 (2008-10-28)
On the basis of high enzyme activity a newly isolated strain of L. mesenteroides CMG713 was selected for dextran production. For maximum yield of dextran, effects of various parameters such as pH, temperature, sucrose concentration and incubation period were studied.
Yang Yang et al.
Biochimica et biophysica acta, 1840(1), 288-293 (2013-10-01)
Abnormal adhesion of red blood cells (RBCs) to vascular endothelium is often associated with reduced levels of sialic acids on RBC membranes and with elevated levels of pro-adhesive plasma proteins. However, the synergistic effects of these two factors on the
Elizabeth M Bradshaw et al.
Nature neuroscience, 16(7), 848-850 (2013-05-28)
In our functional dissection of the CD33 Alzheimer's disease susceptibility locus, we found that the rs3865444(C) risk allele was associated with greater cell surface expression of CD33 in the monocytes (t50 = 10.06, P(joint) = 1.3 × 10(-13)) of young
Hideto Katsuta et al.
Investigative ophthalmology & visual science, 54(6), 4102-4108 (2013-05-23)
To elucidate the role of signals mediated by EphB4 receptor tyrosine kinase and its transmembrane ephrinB2 ligand in corneal lymphatic capillaries. To detect expression of ephrinB2 and EphB4 in mouse corneas, immunohistochemistry of flat-mount corneas from 6- to 10-week-old wild-type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service