Skip to Content
Merck
All Photos(1)

Key Documents

31390

Sigma-Aldrich

Dextran from Leuconostoc spp.

Mr ~70,000

Synonym(s):

DEXTRAN 70

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

(Leuconostoc spp)

form

powder

mol wt

Mr ~70,000

loss

≤7% loss on drying

color

white

mp

483  °C ((901 °F ))

solubility

water: 0.75 g/5mL, clear

absorption

≤0.05 at 375 nm in H2O at 10%

storage temp.

room temp

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dextran is an extracellular bacterial polymer of D-glucopyranose. It primarily consists of α-(1→6) linkage in the main chain and a variable amount of α-(1→2), α-(1→3), α-(1→4) branched linkages.

Application

Dextran from Leuconostoc spp. has been used:
  • for the isolation of islets from pancreatic tissue by centrifugation
  • as a component of Earle′s bicarbonate buffer for placental perfusion
  • for the isolation of rat brain microvessels

Biochem/physiol Actions

Dextran is a, which is commercially used as a drug, particularly as a blood plasma volume expander. It is used as an adjuvant, emulsifier, carrier, and stabilizer in the food, pharmaceutical, and chemical industries. Sephadex, a cross-linked dextran is broadly used for the separation and purification of protein. In the food industry, dextran found its use as a thickener for jam and ice cream. It inhibits the crystallization of sugar, thereby enhancing moisture retention. This helps in maintaining the flavor and appearance of various food items.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.
Enhances immunoprecipitation sensitivity

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sin-Yu Yang et al.
Transplantation proceedings, 51(5), 1458-1462 (2019-06-04)
Subcutaneous pockets provide an extrahepatic transplant site for islet grafting to treat type 1 diabetes. However, a hypoxic environment may cause central necrosis to islets and lead to graft failure. Our previous studies focused on a pre-treated subcutaneous site with
M F Dumon et al.
Electrophoresis, 11(10), 846-851 (1990-10-01)
A modification of crossed immunoelectrophoresis for the analysis of plasma lipoproteins is described and is called polyacrylamide gel-crossed immunoelectrophoresis. The incorporation of albumin in the first-dimensional gel facilitates the transfer of the larger lipoproteins containing apolipoprotein B from the first-dimensional
Farwa Sarwat et al.
International journal of biological sciences, 4(6), 379-386 (2008-10-28)
On the basis of high enzyme activity a newly isolated strain of L. mesenteroides CMG713 was selected for dextran production. For maximum yield of dextran, effects of various parameters such as pH, temperature, sucrose concentration and incubation period were studied.
Reka Natarajan et al.
Scientific reports, 8(1), 10558-10558 (2018-07-14)
Cerebrovascular damage caused by either exposure to stress or the widely abused drug, methamphetamine (Meth) is known but stress and drug abuse frequently occur in tandem that may impact their individual cerebrovascular effects. This study examined their co-morbid cerebrovascular effects
W Michael Babinchak et al.
Nature communications, 11(1), 5574-5574 (2020-11-06)
Liquid-liquid phase separation (LLPS) of proteins that leads to formation of membrane-less organelles is critical to many biochemical processes in the cell. However, dysregulated LLPS can also facilitate aberrant phase transitions and lead to protein aggregation and disease. Accordingly, there

Articles

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service