Skip to Content
Merck
All Photos(1)

Documents

D4522

Sigma-Aldrich

Deoxyribonucleic acid from calf thymus

Type XV, Activated, lyophilized powder

Synonym(s):

Calf Thymus DNA, DNA from calf thymus, Thymonucleic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106310
eCl@ss:
32160414
NACRES:
NA.52

grade

for molecular biology

Quality Level

type

Type XV

form

lyophilized powder

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

High quality double-stranded template DNA isolated from the thymus of male and female calves.

Application

Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.
Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations.

Features and Benefits

• No nuclease activity detected after 16 h at 37 °C incubation
• High quality template DNA

Quantity

One mg DNA is approx. 20 A260 units.

Preparation Note

Prepared from highly polymerized calf thymus DNA and nicked with DNase using the method of Aposhian and Kornberg.

Reconstitution

Dissolve the DNA gently rocking overnight to a final concentration of 1mg/ml in molecular biology grade water.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parul Natvar Patel et al.
Tissue engineering, 11(9-10), 1498-1505 (2005-11-02)
The ultimate goal of this research is to develop an injectable cell-scaffold system capable of permitting adipogenesis to abrogate soft tissue deficiencies resulting from trauma, tumor resection, and congenital abnormalities. The present work compares the efficacy of photopolymerizable poly(ethylene glycol)
Claudia Gomes et al.
Life science alliance, 4(11) (2021-09-11)
High levels of autoimmune antibodies are observed in COVID-19 patients but their specific contribution to disease severity and clinical manifestations remains poorly understood. We performed a retrospective study of 115 COVID-19 hospitalized patients with different degrees of severity to analyze
Juan Antonio Aguilar-Pimentel et al.
Scientific reports, 10(1), 18334-18334 (2020-10-29)
Sex steroids, such as estrogens and androgens, are important regulators of the humoral immune response. Studies in female mice have demonstrated that alteration of circulating estrogen concentration regulates antibody-mediated immunity. As males have normally little endogenous estrogen, we hypothesized that
Brian D Holt et al.
Advanced healthcare materials, 5(23), 3056-3066 (2016-12-08)
Synthetic biomaterials are poised to transform medicine; however, current synthetic options have yet to ideally recapitulate the desirable properties of native tissue. Thus, the development of new synthetic biomaterials remains an active challenge. Due to its excellent properties, including electrical
Anastasia S Singatulina et al.
Cell reports, 27(6), 1809-1821 (2019-05-09)
PARP-1 synthesizes long poly(ADP-ribose) chains (PAR) at DNA damage sites to recruit DNA repair factors. Among proteins relocated on damaged DNA, the RNA-binding protein FUS is one of the most abundant, raising the issue about its involvement in DNA repair.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service