Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
type
Type VIII
Quality Segment
form
fibers
impurities
<2% protein
color
, white to white with yellow cast
solubility
water: 0.98-1.02 mg/mL, faintly hazy to hazy, colorless to light yellow
λmax
256-264 nm (15mM NaCl+1.5mM Trisodium Citrate PH7)
storage temp.
2-8°C
Application
Preparation Note
1 of 1
This Item | |||
|---|---|---|---|
| description Type VIII | description Genomic, unsheared | description Type I, fibers | description - |
| Quality Level 200 | Quality Level 200 | Quality Level 300 | Quality Level 200 |
| form fibers | form lyophilized powder | form fibers | form solid |
| solubility water: 0.98-1.02 mg/mL, faintly hazy to hazy, colorless to light yellow | solubility water: ~1 g/L | solubility - | solubility H2O: 2 mg/mL, clear to hazy |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| color , white to white with yellow cast | color - | color white | color - |
| impurities <2% protein | impurities - | impurities - | impurities - |
Still not finding the right product?
Explore all of our products under Deoxyribonucleic acid sodium salt from Escherichia coli strain B
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



