Skip to Content
Merck
All Photos(1)

Key Documents

D4506

Sigma-Aldrich

4-(Dimethylamino)cinnamaldehyde

>97.5% purity (HPLC), powder

Synonym(s):

DMAC, DMACA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CH=CHCHO
CAS Number:
Molecular Weight:
175.23
Beilstein:
972369
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

4-(Dimethylamino)cinnamaldehyde, chromogenic reagent for indoles and flavanols

Assay

≥98% (HPLC)

form

powder

technique(s)

titration: suitable

color

yellow to very dark yellow

mp

138-140 °C (lit.)

solubility

chloroform/ethanol (1:1): 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

CN(C)c1ccc(\C=C\C=O)cc1

InChI

1S/C11H13NO/c1-12(2)11-7-5-10(6-8-11)4-3-9-13/h3-9H,1-2H3/b4-3+

InChI key

RUKJCCIJLIMGEP-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 4-(Dimethylamino)cinnamaldehyde (DMAC) has been used in the serine protease inhibition assay.
  • It has been used for the detection of (epi)-flavan-3-ols and proanthocyanidin polymers in Brassica napus seeds.
  • It has been used for the determination of phenolic compounds in leaf samples.
  • It has been used for the staining of flavan-3-ols in plant samples.

Biochem/physiol Actions

Reagent for assay of indole product of apotryptophanase and tryptophanase. DMAC produces colored adducts with flavanols for subsequent HPLC. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Analysis of flavanols in beverages by high-performance liquid chromatography with chemical reaction detection.
de Pascual-Teresa, S., et al.
Journal of Agricultural and Food Chemistry, 46, 4209-4213 (1998)
The promoter of the Arabidopsis thaliana BAN gene is active in proanthocyanidin-accumulating cells of the Brassica napus seed coat.
Nesi N, et al.
Plant Cell Reports, 28, 601-601 (2009)
Co-occurrence of tannin and tannin-less vacuoles in sensitive plants.
Fleurat-Lessard P, et al.
Protoplasma, 253, 821-821 (2016)
Exploring plant defense theory in tall goldenrod, Solidago altissima.
Heath JJ, et al.
The New phytologist, 202, 1357-1357 (2014)
A new reagent for the assay of indole in the tryptophanase reaction.
J M TURNER
The Biochemical journal, 78, 790-792 (1961-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service