Skip to Content
Merck
All Photos(1)

Key Documents

D4287

Sigma-Aldrich

γ,γ-Dimethylallyl pyrophosphate triammonium salt

1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥90% (TLC)

Synonym(s):

DMAPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O7P2 · 3NH3
CAS Number:
Molecular Weight:
297.18
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (TLC)

form

liquid

packaging

vial of 200 μg

concentration

1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3))

storage temp.

−20°C

SMILES string

N.N.N.C\C(C)=C\COP(O)(=O)OP(O)(O)=O

InChI

1S/C5H12O7P2.3H3N/c1-5(2)3-4-11-14(9,10)12-13(6,7)8;;;/h3H,4H2,1-2H3,(H,9,10)(H2,6,7,8);3*1H3

InChI key

VBUNGGIXIOHBHL-UHFFFAOYSA-N

Biochem/physiol Actions

Intermediate in terpene biosynthesis

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Yoneyama et al.
Plant & cell physiology, 57(12), 2497-2509 (2016-12-18)
Soybean (Glycine max) accumulates several prenylated isoflavonoid phytoalexins, collectively referred to as glyceollins. Glyceollins (I, II, III, IV and V) possess modified pterocarpan skeletons with C
Carla Araya-Cloutier et al.
PloS one, 12(3), e0174665-e0174665 (2017-03-30)
Acceptor substrate specificity of Streptomyces roseochromogenes prenyltransferase SrCloQ was investigated using different non-genuine phenolic compounds. RP-UHPLC-UV-MSn was used for the tentative annotation and quantification of the prenylated products. Flavonoids, isoflavonoids and stilbenoids with different types of substitution were prenylated by
Xi Zhang et al.
Journal of chromatography. A, 1558, 115-119 (2018-05-19)
The natural rubber molecule is one of the end products of isoprenoids metabolism in the plant. Dimethylallyl diphosphate (DMAPP) and farnesyl pyrophosphate (FPP) are two typical isoprenoids which control the rate of biosynthesis and the molecular weight of natural rubber.
Khiem Nguyen et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(11), 4697-4706 (2017-07-15)
Small isoprenoid diphosphates, such as (
Jinyue Liu et al.
Plant & cell physiology, 59(1), 128-141 (2017-11-16)
Prenylated isoflavonoids have been found in several legume plants, and they possess various biological activities that play important roles in both plant defense and human health. However, it is still unknown whether prenylated isoflavonoids are present in the model legume

Articles

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service