Skip to Content
Merck
All Photos(1)

Documents

78903

Sigma-Aldrich

(E/Z)-Phytoene

mixture of isomers, ≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H64
CAS Number:
Molecular Weight:
544.94
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95% (HPLC)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC\C(C)=C\C=C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(\C)C

InChI

1S/C40H64/c1-33(2)19-13-23-37(7)27-17-31-39(9)29-15-25-35(5)21-11-12-22-36(6)26-16-30-40(10)32-18-28-38(8)24-14-20-34(3)4/h11-12,19-22,27-30H,13-18,23-26,31-32H2,1-10H3/b12-11+,35-21+,36-22+,37-27+,38-28+,39-29+,40-30+

InChI key

YVLPJIGOMTXXLP-KEKOKYSKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phytoene is a colorless carotenoid, that is bioavailable and may be beneficial for health. It belongs to the carotenoid family and can absorb light in the UV (ultra violet) range (λ max 287nm).

Application

(E/Z)-Phytoene has been used as a reference standard to prepare the standard curve in high-performance liquid chromatography (HPLC).

Biochem/physiol Actions

Phytoene acts as precursors for other carotenoids in nature. It can also serve as free radical quenchers, DNA protectors and anti-inflammatory agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The photoprotective effects of a food supplement tomato powder rich in phytoene and phytofluene, the colorless carotenoids, a preliminary study
von O B L, et al.
Global Journal of Research In Engineering, 2, 178-182 (2015)
Accumulation of phytoene, a colorless carotenoid by inhibition of phytoene desaturase (PDS) gene in Dunaliella salina V-101
Srinivasan R, et al.
Bioresource Technology, 242, 311-318 (2017)
Klaudia Ciurkot et al.
Nucleic acids research, 49(13), 7775-7790 (2021-07-02)
CRISPR Cas12a is an RNA-programmable endonuclease particularly suitable for gene regulation. This is due to its preference for T-rich PAMs that allows it to more easily target AT-rich promoter sequences, and built-in RNase activity which can process a single CRISPR
The colourless carotenoids phytoene and phytofluene: From dietary sources to their usefulness for the functional foods and nutricosmetics industries
Melendez-Martinez A J, et al.
J. Food Compos. Anal., 67, 91-103 (2018)
Kelly Laje et al.
Marine drugs, 17(3) (2019-03-27)
Carotenoids are lipophilic pigments found in plants and algae, as well as some bacteria, archaea, and fungi that serve two functions-(1) as light harvesting molecules-primary carotenoids, and (2) as antioxidants, acting against reactive oxygen species⁻secondary carotenoids. Because of their strong

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service