Skip to Content
Merck
All Photos(1)

Key Documents

D1377

Sigma-Aldrich

2,6-Diaminopimelic acid

≥98% (TLC), suitable for thin layer chromatography (TLC)

Synonym(s):

2,6-Diaminoheptanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(NH2)(CH2)3CH(NH2)COOH
CAS Number:
Molecular Weight:
190.20
Beilstein:
1787719
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

2,6-Diaminopimelic acid, ≥98% (TLC)

Assay

≥98% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

color

white to off-white

mp

~295 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NC(CCCC(N)C(O)=O)C(O)=O

InChI

1S/C7H14N2O4/c8-4(6(10)11)2-1-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)

InChI key

GMKMEZVLHJARHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Diaminopimelic acid is produced in bacteria and is a component of cell wall. It is also synthesized by higher plants.

Application

2,6-Diaminopimelic acid (DAP) has been used:
  • as a supplement in glucose minimal media for screening Bacillus subtilis(44)
  • as a component of lysogeny broth (LB) agar for screening E coli transformants(45)
  • as a standard in thin layer chromatography for the characterization of DAP isoforms from Streptomyces strains(46)

Biochem/physiol Actions

2,6-Diaminopimelic acid is essential for microbial metabolism and is a potential marker for quantification of microbial proteins. It is a sensitive and reliable marker and is majorly quantified using gradient high-performance liquid chromatography (HPLC).

Other Notes

Mixture of LL-, DD- and meso-isomers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Construction and Analysis of Two Genome-Scale Deletion Libraries for Bacillus subtilis
Byoung-MoKoo
Cell Systems, 4, 291-305 (2017)
Determination of 2, 6-diaminopimelic acid in bacteria, ruminal and duodenal digesta using HPLC with fluorescence or UV detection
Czauderna M and Kowalczyk J
Journal of Animal and feed sciences, 8(5), 273-288 (1999)
The Structure and Function of the Membranes and Surfaces of Cells, 22, 77-77 (1963)
2, 6-Diaminopimelic acid (DAPA) in microbial protein quantification of heifers fed different forage sources
Couto Filho CCC, et al.
Revista Brasileira de Zootecnia, 44(5), 180-186 (2015)
B R Hamaker et al.
Journal of pediatric gastroenterology and nutrition, 13(1), 59-66 (1991-07-01)
Metabolic balance studies were carried out in young children fed diets based on maize, amaranth, or cassava flours and in corresponding casein controls. Dietary fiber intakes were 22.2, 20.5, and 9.0 g/day for the maize, amaranth, and cassava groups, respectively.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service