Skip to Content
Merck
All Photos(1)

Key Documents

D1319

Sigma-Aldrich

Anti-DAP Kinase 1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DAPK, Anti-DAPK1, Anti-Death associated protein kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~160 kDa

species reactivity

mouse (predicted), human, rat (predicted)

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5.0 μg using extracts of human HeLa cells
western blot: 1-2 μg/mL using whole extracts of human HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DAPK1(1612)
mouse ... Dapk1(69635)
rat ... Dapk1(306722)

Related Categories

General description

DAP-kinase 1 is a Ca2+/calmodulin dependent Ser/Thr kinase that associates with microfilaments. The protein is composed of a multidomain structure. It has a subdomain typical of serine/threonine kinases, a Ca2+/calmodulin regulatory domain, eight ankyrin repeats followed by two P-loop motifs and a typical death domain module. It contains two auto-inhibitory domains, one being Ca2+/calmodulin dependent.

Specificity

Anti-DAPK1 antibody is specific for human DAPK1 (approx. 160 kDa). In immunoblotting, detection of the DAPK1 band is specifically inhibited with the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 2-13 of human DAP-kinase 1, conjugated to KLH via a C-terminal cysteine residue. The corresponding sequence is identical in rat and mouse.

Application

Anti-DAP Kinase 1 antibody is suitable for use in immunoprecipitation (2.5-5.0 μg using extracts of human HeLa cells) and western blot (1-2 μg/mL using whole extracts of human HeLa cells).
Anti-DAP Kinase 1 antibody produced in rabbit has been used in immunoprecipitation and western blotting.

Biochem/physiol Actions

Death-associated protein kinase (DAPK) belongs to the serine/threonine kinase family that regulates apoptosis, growth factor signaling and also may be involved in modulating immune responses. DAPK1 mediates cellular functions such as membrane blebbing and autophagy and inhibits the activity of prolyl isomerase

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grifolin, a potent antitumour natural product upregulates death-associated protein kinase 1 DAPK1 via p53 in nasopharyngeal carcinoma cells
Luo XJ, et al.
European Journal of Cancer, 47(2), 316-325 (2011)
Yao Lin et al.
The FEBS journal, 277(1), 48-57 (2009-11-03)
Death-associated protein kinase (DAPK) is a stress-regulated protein kinase that mediates a range of processes, including signal-induced cell death and autophagy. Although the kinase domain of DAPK has a range of substrates that mediate its signalling, the additional protein interaction
Iron-loaded transferrin (Tf) is detrimental whereas iron-free Tf confers protection against brain ischemia by modifying blood Tf saturation and subsequent neuronal damage
DeGregorio-RN, et al.
Redox Biology, 143-158 (2018)
DAP-kinase is a Ca2+/calmodulin-dependent, cytoskeletal-associated protein kinase, with cell death-inducing functions that depend on its catalytic activity
Cohen O, et al.
The Embo Journal, 16(5), 998-1008 (1997)
DAPK1 mediates the G1 phase arrest in human nasopharyngeal carcinoma cells induced by grifolin, a potential antitumor natural product
Luo XJ, et al.
European Journal of Pharmacology, 670(2-3), 427-434 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service