Skip to Content
Merck
All Photos(1)

Key Documents

D9656

Sigma-Aldrich

Anti-DNP antibody produced in rabbit

whole antiserum

Synonym(s):

Anti Dnp, Anti Dnp Antibody, Anti-DNP - Anti-DNP antibody produced in rabbit, Anti-Dnp, Anti-Dnp Antibody, Dnp Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:8,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

A DNP-BSA conjugate

Application

Anti-DNP antibody produced in rabbit was used to tag derivatized protein carbonyl groups in plasma by ELISA. It was used for differential cell count of zona-free blastocysts in IVF embryos.

Biochem/physiol Actions

2,4-Dinitrophenyl is an oxidative phosphorylation inhibitor and hinders adenosine triphosphate (ATP) synthesis. Due to its low excretion rate, 2,4-dinitrophenyl is considered toxic to human. It increases electron transportation, respiration and overall metabolism.

Analysis Note

Antiserum is tested for specificity and titer by immunoelectrophoresis; treated to remove lipoproteins..

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jens Nagel et al.
Chromosoma, 121(4), 353-367 (2012-03-15)
The large-scale chromatin organization of retrovirus and retroviral gene vector integration loci has attracted little attention so far. We compared the nuclear organization of transcribed integration loci with the corresponding loci on the homologous chromosomes. Loci containing gamma-retroviral gene transfer
David Wunder et al.
Antimicrobial agents and chemotherapy, 48(1), 110-115 (2003-12-25)
Salivary histatins (Hsts) are potent candidacidal proteins that induce a nonlytic form of cell death in Candida albicans accompanied by loss of mean cell volume, cell cycle arrest, and elevation of intracellular levels of reactive oxygen species (ROS). Since these
Liliana Matos et al.
Oxidative medicine and cellular longevity, 2017, 3793817-3793817 (2017-03-11)
Copper sulfate-induced premature senescence (CuSO
Felipe C G Reis et al.
Aging, 8(6), 1201-1222 (2016-06-01)
Aging increases the risk of type 2 diabetes, and this can be prevented by dietary restriction (DR). We have previously shown that DR inhibits the downregulation of miRNAs and their processing enzymes - mainly Dicer - that occurs with aging
Åsa Fredriksson et al.
Aging cell, 11(4), 634-643 (2012-04-18)
In organisms with a soma-germ demarcation, the germline must be 'preserved' such that harmful damage is not transmitted to the offspring. Keeping the progeny free of damage may be achieved by gametes enjoying elevated, and/or more functional, homeostatic maintenance systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service