Skip to Content
Merck
All Photos(2)

Key Documents

D1306

Sigma-Aldrich

Debrisoquine sulfate

powder, ≥98% (TLC)

Synonym(s):

3,4-Dihydro-2(1H)-isoquinolinecarboximidamide, Debrisoquin sulfate, Ro 5-33071

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H26N6 · H2SO4
CAS Number:
Molecular Weight:
448.54
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Debrisoquine sulfate,

Assay

≥98% (TLC)

Quality Level

form

powder

mp

285 °C

solubility

H2O: 20 mg/mL (with heat)

storage temp.

room temp

SMILES string

OS(O)(=O)=O.NC(=N)N1CCc2ccccc2C1.NC(=N)N3CCc4ccccc4C3

InChI

1S/2C10H13N3.H2O4S/c2*11-10(12)13-6-5-8-3-1-2-4-9(8)7-13;1-5(2,3)4/h2*1-4H,5-7H2,(H3,11,12);(H2,1,2,3,4)

InChI key

CAYGYVYWRIHZCQ-UHFFFAOYSA-N

Gene Information

human ... SLC6A2(6530)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Debrisoquine is an anti-hypertensive agent. It is metabolized by cytochrome P4502D6.

Substrates

A substrate for cytochrome P450 CYP2D6; an indicator for genetic polymorphism in cytochrome P450.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Jacqz-Aigrain et al.
Biochemical pharmacology, 41(11), 1657-1663 (1991-06-01)
Interindividual variations of debrisoquine metabolism was recently identified in non-human primates tested in vivo. The catalytical and immunological characterization of cytochrome P450IID subfamily was undertaken in hepatic microsomes from extensive metabolizer primates. The NADPH/O2 mediated metabolism of debrisoquine, dextromethorphan and
N E Caporaso et al.
Environmental health perspectives, 98, 101-105 (1992-11-01)
Debrisoquine is an antihypertensive drug that is metabolized by cytochrome P4502D6. Deficient metabolism is inherited as an autosomal recessive condition. We previously reported in a case-control study that extensive metabolizers of debrisoquine were at greater risk of lung cancer compared
Ping-Ching Hsu et al.
Molecular carcinogenesis, 56(2), 594-606 (2016-06-25)
Smoking-related biomarkers for lung cancer and other diseases are needed to enhance early detection strategies and to provide a science base for tobacco product regulation. An untargeted metabolomics approach by ultra-performance liquid chromatography-quadrupole-time of flight mass spectrometry (UHPLC-Q-TOF MS) totaling
J W Ho et al.
Analytical biochemistry, 203(2), 348-351 (1992-06-01)
Debrisoquine and sparteine are prototype substrates of a genetic deficiency in cytochrome P450-dependent drug metabolism. Sensitive assays of in vitro oxidation of sparteine and debrisoquine are required for evaluation of this polymorphism. The activities were measured by quantitative analysis of
E Scarr et al.
Progress in neuro-psychopharmacology & biological psychiatry, 15(2), 297-301 (1991-01-01)
1. Studies were carried out on three monoamine oxidase (MAO) inhibitors, two of which, debrisoquine and para- hydroxyphenelzine, are purported to be peripheral inhibitors and one, phenelzine, is a peripherally acting inhibitor, which has been included for comparitive purposes. 2.

Articles

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service