Skip to Content
Merck
All Photos(1)

Key Documents

D1283

Sigma-Aldrich

Dulbecco′s Phosphate Buffered Saline

With calcium chloride and magnesium chloride, 10×, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DPBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
eCl@ss:
32129211
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

glucose: no (Dextro)
sodium pyruvate: no

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Phosphate Buffered Saline is an isotonic saline solution used for washing cells and tissues. This balanced salt solution (BSS) is also used in cell culture as a buffering system to maintain the medium within the physiological pH range and as an irrigating, transporting and diluting fluid while maintaining intra- and extracellular osmotic balance.

Application

Dulbecco′s Phosphate Buffered Saline has been used:
  • for decellularization of extracellular matrix (ECM)
  • to rinse biosensor chip to remove physically adsorbed mycolic acids
  • for washing cells before staining during in vitro biocompatibility test

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nasim Shahidi Hamedani et al.
Journal of clinical medicine, 10(4) (2021-02-14)
Coagulation factor XIII (FXIII) is a protransglutaminase which plays an important role in clot stabilization and composition by cross-linking the α- and γ-chains of fibrin and increasing the resistance of the clot to mechanical and proteolytic challenges. In this study
Sarah S Gabriel et al.
Kidney international, 91(4), 842-855 (2017-02-02)
Dent disease is a rare X-linked tubulopathy caused by mutations in the endosomal chloride-proton exchanger (ClC-5) resulting in defective receptor-mediated endocytosis and severe proximal tubule dysfunction. Bone marrow transplantation has recently been shown to preserve kidney function in cystinosis, a
Alba Di Pardo et al.
Cell death & disease, 11(7), 546-546 (2020-07-20)
The longevity-associated variant (LAV) of the bactericidal/permeability-increasing fold-containing family B member 4 (BPIFB4) has been found significantly enriched in long-living individuals. Neuroinflammation is a key player in Huntington's disease (HD), a neurodegenerative disorder caused by neural death due to expanded
Ho-Ying Wan et al.
Acta biomaterialia, 140, 163-177 (2021-12-08)
The development of bone-like tissues in vitro that exhibit key features similar to those in vivo is needed to produce tissue models for drug screening and the study of bone physiology and disease pathogenesis. Extracellular matrix (ECM) is a predominant
Janice Dias et al.
Eye & contact lens, 41(5), 281-286 (2015-01-21)
To determine the effect of hydration media on ex vivo corneal elasticity. Experiments were conducted on 40 porcine eyes retrieved from an abattoir (10 eyes each for phosphate-buffered saline (PBS), balanced salt solution, Optisol, 15% dextran). The epithelium was removed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service