Skip to Content
Merck
All Photos(1)

Key Documents

11666789001

Roche

Buffers in a Box, Premixed PBS Buffer, 10x

10X, Premixed, solurion (clear colorless)

Synonym(s):

PBS buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Product Name

Buffers in a Box, Premixed PBS Buffer, 10x, solution (clear, colorless), optimum pH ~7.0

form

solution (clear, colorless)

packaging

pkg of 4 L

manufacturer/tradename

Roche

concentration

10 ×

optimum pH

~7.0

shipped in

ambient

storage temp.

20-25°C

General description

The premixed PBS (phosphate buffered saline) buffer is provided as a 10x solution.

Application

Buffers in a Box, Premixed PBS Buffer, 10x has been used in immunohistochemistry and fluorescence in situ hybridization.
Buffers in a Box, Premixed PBS Buffer, 10x has been used:
  • to dissolve Candida albicans to prepare its inoculum
  • in western blotting assay to wash the poly(vinylidene fluoride) (PVDF) membranes incubated with horseradish peroxidase (HRP)-conjugated secondary antibodies
  • for resuspending pellets for testing treated dogs′ sera for antibodies to Islet cells and adipose-derived stem cells (ASCs)

Analysis Note

Absence of contaminants: Free of DNase and RNase according to the current Quality Control procedures.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oluwaseun Adebayo Bamodu et al.
Cancers, 13(16) (2021-08-28)
prostate cancer (PCa) is a principal cause of cancer-related morbidity and mortality. Castration resistance and metastasis are clinical challenges and continue to impede therapeutic success, despite diagnostic and therapeutic advances. There are reports of the oncogenic activity of genetic suppressor
Anna Gooch et al.
PloS one, 14(9), e0218688-e0218688 (2019-09-20)
We previously reported that allogeneic, intraperitoneally administered "Neo-Islets," composed of cultured pancreatic islet cells co-aggregated with high numbers of immunoprotective and cytoprotective Adipose-derived Stem Cells, reestablished, through omental engraftment, redifferentiation and splenic and omental up-regulation of regulatory T-cells, normoglycemia in
Tracy D Ainsworth et al.
The ISME journal, 9(10), 2261-2274 (2015-04-18)
Despite being one of the simplest metazoans, corals harbor some of the most highly diverse and abundant microbial communities. Differentiating core, symbiotic bacteria from this diverse host-associated consortium is essential for characterizing the functional contributions of bacteria but has not
Wayan Larissa Demolsky et al.
European journal of dentistry (2021-12-23)
Oral candidiasis is an infection caused by pathogenic fungi Candida albicans, with a considerably high prevalence of 20 to 72%. Indian camphorweed (Pluchea indica) also known as "beluntas" as the local name has been known as a traditional medicine in
Oluwaseun Adebayo Bamodu et al.
Cancers, 13(16) (2021-08-28)
prostate cancer (PCa) is a principal cause of cancer-related morbidity and mortality. Castration resistance and metastasis are clinical challenges and continue to impede therapeutic success, despite diagnostic and therapeutic advances. There are reports of the oncogenic activity of genetic suppressor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service