D0672
φX174 DNA Marker Hae III Digest
for DNA electrophoresis
About This Item
Recommended Products
Quality Level
100
200
form
liquid
usage
200 uses (0.1 mg size)
40 uses (20 μg size)
suitability
suitable for electrophoresis (DNA)
storage temp.
−20°C
General description
Components
Preparation Note
Other Notes
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.
Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.
Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.
Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service