Skip to Content
Merck
All Photos(1)

Key Documents

D7058

Sigma-Aldrich

DirectLoad Wide Range DNA Marker

ready-to-use marker for DNA electrophoresis

Synonym(s):

DirectLoad marker, RTU DNA ladder, RTU DNA marker, agarose gel electrophoresis ladder, agarose gel electrophoresis marker, gel ladder, gel marker, wide range ladder, wide range ladder for gel electrophoresis, wide range marker for gel electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105335
NACRES:
NA.25

Quality Level

form

liquid

usage

100 uses

suitability

suitable for electrophoresis (DNA)

storage temp.

−20°C

General description

Sigma′s DirectLoad Wide Range DNA Marker contains 16 fragments from 50 – 10,000 bp, with bands regularly spaced. Ten ul of the ladder can be loaded directly in a single lane on an agarose or polyacrylamide gel.

Application

Suitable for size determination of PCR-generated DNA fragments with either agarose or polyacrylamide gels.

Features and Benefits

  • Ready-to-load
  • Easy-to-use
  • Popular band sizes

Components

Sigma′s DirectLoad Wide Range DNA Marker is provided in a gel-loading solution of 2.5% Ficoll (Type 400), 0.0125% bromophenol blue, and 0.00625% xylene cyanol.

Other Notes

Contains 16 fragments, 50–10,000 bp which produce logically spaced bands on an agarose gel.
For optimal resolution, the recommended agarose gel concentration is 0.75 – 2.0%.

Legal Information

DirectLoad is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melanie P Dautle et al.
Journal of clinical microbiology, 40(2), 414-421 (2002-02-05)
In this study, 83 clinical isolates purified from biofilms colonizing 18 silicone gastrostomy devices (12 "buttons" and six tubes converted to skin level devices) were selected for subtype characterization utilizing genetic analysis. The tubes, previously used for feeding, remained in

Articles

Colony PCR reagents and our colony PCR protocol using REDExtract-N-Amp™ PCR ReadyMix™ and JumpStart™ REDTaq® PCR ReadyMix™ reagents.

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

SeqPlex™-I WTA kit amplifies RNA for NGS, enabling genomic studies from limited samples.

Markers for gel electrophoresis aid size determination of DNA, PCR fragments, and RNA, staining well with common nucleic acid stains.

See All

Protocols

Protocol for high fidelity amplification of long PCR fragments up to 22kb from complex DNA mixtures and up to 40kb from simple DNA mixtures.

Protocol for high fidelity amplification of long PCR fragments up to 22kb from complex DNA mixtures and up to 40kb from simple DNA mixtures.

Protocol for high fidelity amplification of long PCR fragments up to 22kb from complex DNA mixtures and up to 40kb from simple DNA mixtures.

Protocol for high fidelity amplification of long PCR fragments up to 22kb from complex DNA mixtures and up to 40kb from simple DNA mixtures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service