Skip to Content
Merck
All Photos(1)

Key Documents

CLS3458

Corning® Transwell® 24 well plates

8.0 μm pore size round wells, clear wells, clear polystyrene (plate or dish), polycarbonate membrane, sterile, pack of 12, case of 24

Synonym(s):

cell culture inserts, cell culture plate, cell culture plates, corning plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.13

Product Name

Corning® Transwell® polycarbonate membrane cell culture inserts, 6.5 mm Transwell Inserts with 8.0 μm pore polycarbonate membrane coated w/ Cultrex BME, w/lid, sterile, 24/cs

material

clear polystyrene (plate or dish)
clear wells
polycarbonate membrane
round wells

sterility

sterile

feature

lid

packaging

pack of 12
case of 24

manufacturer/tradename

Corning 3458

availability

not available in EU

technique(s)

cell culture | mammalian: suitable

insert working volume

0.1 mL

membrane diam.

6.5 mm

size

24 wells

surface area

0.33 cm2 , cell growth area

well working volume

0.6 mL

color

translucent

pore size

8.0 μm

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • 10 μm thick translucent polycarbonate membrane
  • Packaged 12 inserts in a 24 well plate, 2 plates per case
  • Membrane must be stained for cell visibility
  • Aseptically coated with Cultrex Basement Membrane Extract (BME), a soluble form of extracellular matrix purified from EHS turmors.
  • Ideal for cell invasion experiments
FOR U.S. CUSTOMERS ONLY
Transwell cell culture inserts are convenient, easy-to-use permeable support devices for the study of both anchorage-dependent and anchorage-independent cell lines. They feature a 10 μm thin transparent polycarbonate membrane available in a variety of pore sizes. Transwell are treated for optimal cell attachment. Cells must be stained for cell visibility. The PC membrane is compatible with most organic fixatives and stains.

Legal Information

Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria Vinader et al.
PloS one, 8(10), e78744-e78744 (2013-11-10)
Amongst the chemokine signalling axes involved in cancer, chemokine CXCL12 acting on chemokine receptor CXCR4 is particularly significant since it orchestrates migration of cancer cells in a tissue-specific metastatic process. High CXCR4 tumour expression is associated with poor prognosis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service