Skip to Content
Merck
All Photos(4)

Documents

PICM01250

Millipore

Millicell® Standing Cell Culture Inserts

pore size 0.4 μm, diam. 12 mm, transparent PTFE membrane, hydrophilic, size 24 wells, sterile

Synonym(s):

Millicell Cell Culture Insert, 12 mm, hydrophilic PTFE, 0.4 µm, Millicell-CM, cell culture inserts, permeable culture insert, plate inserts, tissue culture insert, tissue culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22

material

polystyrene housing
transparent PTFE membrane

Quality Level

sterility

ethylene oxide treated
sterile

feature

hydrophilic

packaging

pack of 50

manufacturer/tradename

Millicell®

parameter

50 °C max. temp.

technique(s)

cell attachment: suitable
cell culture | mammalian: suitable
cell differentiation: suitable

H

10.5 mm

diam.

12 mm

filtration area

0.6 cm2

size

24 wells

surface area

0.6 cm2

working volume

0.6 mL

color

transparent, when wetted

matrix

Biopore

pore size

0.4 μm pore size

binding type

low binding surface

detection method

fluorometric

shipped in

ambient

General description

Device Configuration: 24-well plate, 0.4μm pore size

With Millicell® inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell® inserts make it possible to study both sides of the cell monolayer.

Millicell® Standing Inserts:
- Promotes excellent cell growth and provides an exceptional opportunity for cell studies

Membrane Type:
Biopore Membrane (hydrophilic PTFE)
- For low protein-binding, live cell viewing, and immunofluorescent applications

Applications
Cell Attachment, Cell Growth, Cell Differentiation , Immunocytochemistry

Application

Research Category
Cell Culture

Packaging

Individually blister packaged

Legal Information

BIOPORE is a trademark of Merck KGaA, Darmstadt, Germany
Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tong-Dian Zhang et al.
Oxidative medicine and cellular longevity, 2020, 4569268-4569268 (2020-06-23)
Mono-(2-ethylhexyl) phthalate (MEHP) and genistein have been classified as endocrine-disrupting chemicals (EDCs) which interfere with the differentiation and development of the male reproductive system. However, how these two EDCs would affect fetal rat testis development at a low dose was
Gravity in mammalian organ development: differentiation of cultured lung and pancreas rudiments during spaceflight.
Spooner, B S, et al.
The Journal of Experimental Zoology, 269, 212-222 (1994)
Ming Zeng et al.
Gene, 753, 144810-144810 (2020-05-30)
Meiotic entry and progression require dynamic regulation of germline gene expression. m6A on mRNAs and recognition by YTHDC2 has been known as post-transcriptional regulatory complex, but the roles of this regulator remain unclear for meiotic initiation and progression in female
Katerina Jirsova et al.
PloS one, 15(4), e0231473-e0231473 (2020-04-22)
The aim of this study was to determine the effect of autologous serum (AS) eye drops on the density of human leucocyte antigen (HLA)-DR-positive epithelial cells and Langerhans cells on the ocular surface of patients with bilateral severe dry eye
Andrea S Cupp et al.
Biology of reproduction, 68(6), 2033-2037 (2003-02-28)
The first morphological event after initiation of male sex determination is seminiferous cord formation in the embryonic testis. Cord formation requires migration of pre-peritubular myoid cells from the adjacent mesonephros. The embryonic Sertoli cells are the first testicular cells to

Articles

16HBE14o- human bronchial epithelial cells used to model respiratory epithelium for the research of cystic fibrosis, viral pulmonary pathology (SARS-CoV), asthma, COPD, effects of smoking and air pollution. See over 5k publications.

16HBE14o- human bronchial epithelial cells used to model respiratory epithelium for the research of cystic fibrosis, viral pulmonary pathology (SARS-CoV), asthma, COPD, effects of smoking and air pollution. See over 5k publications.

Protocols

Toluidine blue selectively stains nuclear material and acidic tissue components, aiding in histological staining for tissues rich in DNA/RNA.

Toluidine blue selectively stains nuclear material and acidic tissue components, aiding in histological staining for tissues rich in DNA/RNA.

Toluidine blue selectively stains nuclear material and acidic tissue components, aiding in histological staining for tissues rich in DNA/RNA.

Toluidine blue selectively stains nuclear material and acidic tissue components, aiding in histological staining for tissues rich in DNA/RNA.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service