Detroit 551
86082802, human skin (embryonic), Fibroblast
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
Detroit 551, 86082802
biological source
human skin (embryonic)
growth mode
Adherent
karyotype
2n = 46
morphology
Fibroblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian embryo skin
Cell Line Description
Derived from a skin biopsy of a normal Caucasian female embryo. Cells may be considered normal and may serve as controls for study of cells having mutant chromosomes. Cells support growth of herpes viruses and SV40 virus can induce transformation.
Application
Chromosome and virus studies (Normal control): herpesviruses, SV40
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10,12
D13S317: 11,12
D16S539: 12,13
D5S818: 12,13
D7S820: 10,12
THO1: 9,9.3
TPOX: 8
vWA: 18
CSF1PO: 10,12
D13S317: 11,12
D16S539: 12,13
D5S818: 12,13
D7S820: 10,12
THO1: 9,9.3
TPOX: 8
vWA: 18
Culture Medium
EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:5 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Finite lifespan; at most 25 subcultures from original tissue.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service