Skip to Content
Merck
All Photos(1)

Key Documents

C7767

Sigma-Aldrich

Cottonseed oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

plant (Gossypium hirsutum L.)

form

liquid

composition

unsaturated fatty acids, ~70%

density

0.92 g/mL at 25 °C (lit.)

lipid type

oils

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cottonseed oil is extracted from the kernel, a part of the cotton seed. This oil contains linoleic acid, oleic acid, linolenic acid and myristic acid. It is used as non-edible oil and is used in the preparation of polymeric resins.
Cottonseed oil is extracted from the seeds of Gossypium sp. It is used to impart a roasted or nutty aroma to fried food products. Cottonseed oil is used as a component to formulate shortenings and margarines. It has a long shelf life and is healthy for the heart.

Application

Cottonseed oil has been used as an nutritional supplement for alveolar septal wall thickness in mice. It has been used to study its effects on indirect gestational and lactational exposure in rats.
Cottonseed oil has been used to dissolve testosterone propionate employed in preparation of prenatal testosterone -treated Suffolk ewes.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Technological Innovations in Major World Oil Crops, Volume 1: Breeding (2011)
Vegetable Oils in Food Technology: Composition, Properties and Uses (2011)
Modelling bronchopulmonary dysplasia in mice: how much oxygen is enough?
Nardiello C, et al.
Disease models & mechanisms, dmm-027086 (2016)
Virender K Rehan et al.
American journal of physiology. Lung cellular and molecular physiology, 299(5), L672-L680 (2010-08-24)
The physiological development and homeostasis of the lung alveolus is determined by the expression of peroxisome proliferator-activated receptor-γ (PPAR-γ) by the interstitial lipofibroblast. We have recently shown (Dasgupta C et al., Am J Physiol Lung Cell Mol Physiol 296: L1031-L1041
Eco-friendly polyurethane coatings from cottonseed and karanja oil
Gaikwad MS, et al.
Progress in Organic Coatings, 86, 164-172 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service