Skip to Content
Merck
All Photos(1)

Key Documents

C7020

Supelco

Cefadroxil

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O5S
CAS Number:
Molecular Weight:
363.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

form

powder or crystals

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)c3ccc(O)cc3)C2=O)C(O)=O

InChI

1S/C16H17N3O5S/c1-7-6-25-15-11(14(22)19(15)12(7)16(23)24)18-13(21)10(17)8-2-4-9(20)5-3-8/h2-5,10-11,15,20H,6,17H2,1H3,(H,18,21)(H,23,24)/t10-,11-,15-/m1/s1

InChI key

BOEGTKLJZSQCCD-UEKVPHQBSA-N

General description

Cefadroxil is a broad spectrum cephalosporin antibiotic, widely used in veterinery medicine for the oral treatment of infectious diseases.

Application

Cefadroxil has been used as a standard in the analysis of cefadroxil residues in the samples of domestic sheep milk using penzym enzymatic test method. Cefadroxil may be used as an analytical reference standard for the quantification of the analyte in milk samples using Delvotest photometric technique and in biological samples using high-performance liquid chromatography, respectively.
Cefadroxil has been used as a standard in the analysis of cefadroxil residues in the samples of domestic sheep milk using penzym enzymatic test method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Lamba et al.
The West Indian medical journal, 60(3), 346-348 (2012-01-10)
We report a life-threatening anaphylactic reaction to cefadroxil in a 60-year old female with no previous history of allergies to penicillins. Cefadroxil is a first-generation cephalosporin and anaphylactic reactions to it in patients with no previous history of penicillin allergy
Robert Six et al.
Journal of the American Veterinary Medical Association, 233(3), 433-439 (2008-08-05)
To evaluate the efficacy and safety of administration of cefovecin, compared with cefadroxil, for treatment of naturally occurring secondary superficial pyoderma, abscesses, and infected wounds in dogs. Multicenter, randomized, positive-controlled clinical trial. 235 client-owned dogs. Dogs with clinical signs of
Eunice Kazue Kano et al.
Arzneimittel-Forschung, 58(1), 42-47 (2008-03-29)
Two different cefadroxil (CAS 50370-12-2) formulations were evaluated for their relative bioavailability in 24 healthy volunteers who received a single 500 mg oral dose of each preparation. An open, randomized clinical trial designed as a two-period crossover study with a
Scleredema following scabies infestation.
Amit K Malhotra et al.
Pediatric dermatology, 25(1), 136-138 (2008-02-29)
DRESS syndrome from cefadroxil confirmed by positive patch test.
M Hernanto et al.
Allergy, 62(10), 1216-1217 (2007-06-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service