Skip to Content
Merck
All Photos(1)

Key Documents

C5145

Sigma-Aldrich

Chlormadinone acetate

≥98%

Synonym(s):

17α-Acetoxy-6-chloro-4,6-pregnadiene-3,20-dione, 6-Chloro-17α-hydroxy-4,6-pregnadiene-3,20-dione 17-acetate, 6-Chloro-6-dehydro-17α-acetoxyprogesterone, Gestafortin, Matrol, Menstridyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29ClO4
CAS Number:
Molecular Weight:
404.93
Beilstein:
2633614
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

powder

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@]3(OC(C)=O)C(C)=O)[C@]1([H])C=C(Cl)C4=CC(=O)CC[C@]24C

InChI

1S/C23H29ClO4/c1-13(25)23(28-14(2)26)10-7-18-16-12-20(24)19-11-15(27)5-8-21(19,3)17(16)6-9-22(18,23)4/h11-12,16-18H,5-10H2,1-4H3/t16-,17+,18+,21-,22+,23+/m1/s1

InChI key

QMBJSIBWORFWQT-DFXBJWIESA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Review of progestogenic and related pharmacological effects of chlormadinone acetate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathalie Chabbert-Buffet et al.
Contraception, 83(3), 229-237 (2011-02-12)
Systemic lupus erythematosus (SLE) affects women of child-bearing age. Combined oral contraceptives can worsen the course and increase the risk of thrombosis. The objectives of this study were to provide an alternative contraception and thus evaluate the gynecological tolerability of
Ting Huang et al.
Die Pharmazie, 66(3), 212-215 (2011-05-11)
Chlormadinone acetate (CMA), a derivative of 17-a-hydroxyprogesterone, has been widely used as an orally effective progestogen in hormone replacement therapy (HRT). Glucuronidation catalyzed by UDP-glucuronosyltransferases (UGTs) is one of the major steps responsible for the metabolism of many drugs, environmental
Gunther Göretzlehner et al.
Clinical drug investigation, 31(4), 269-277 (2011-01-22)
The prescribing of extended regimens of oral contraceptives (OCs) is increasing in routine gynaecological practice as a means of reducing the number of annual menstrual bleeds. Typically, this involves taking one pill per day for, say, 84 days continuously (4×21
R Hermes et al.
Theriogenology, 78(6), 1217-1223 (2012-08-18)
The estrous cycle length in the white rhinoceros (Ceratotherium simum) is either 4 or 10 wk. The cause(s) for this variation as well as the poor fertility rate in captivity remains under debate in this species. Most captive adult white
Maurizio Guido et al.
Reproductive sciences (Thousand Oaks, Calif.), 17(8), 767-775 (2010-07-03)
This is the first study evaluating the clinical, metabolic, and hormonal effects of the ethinylestradiol-chlormadinone acetate (EECMA) combination in hirsute women with polycystic ovary syndrome (PCOS). Ultrasonographic pelvic examination, hirsutism score, and hormone profile evaluation were performed at baseline and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service