Skip to Content
Merck
All Photos(1)

Key Documents

46698

Supelco

6-Propyl-2-thiouracil

VETRANAL®, analytical standard

Synonym(s):

2,3-Dihydro-6-propyl-2-thioxo-4(1H)-pyrimidinone, 4-Hydroxy-2-mercapto-6-propylpyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2OS
CAS Number:
Molecular Weight:
170.23
Beilstein:
130039
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

enzyme inhibitor

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

218-220 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

CCCC1=CC(=O)NC(=S)N1

InChI

1S/C7H10N2OS/c1-2-3-5-4-6(10)9-7(11)8-5/h4H,2-3H2,1H3,(H2,8,9,10,11)

InChI key

KNAHARQHSZJURB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah V Lipchock et al.
The American journal of clinical nutrition, 98(4), 1136-1143 (2013-09-13)
Alleles of the receptor gene TAS2R38 are responsible in part for the variation in bitter taste perception of 6-n-propylthiouracil (PROP) and structurally similar compounds (eg, glucosinolates in cruciferous vegetables). At low concentrations, people with the PAV ("taster" amino acid sequence)
Michael H Powell et al.
Neurotoxicology, 33(5), 1322-1329 (2012-09-15)
Humans and wildlife are exposed to environmental pollutants that have been shown to interfere with the thyroid hormone system and thus may affect brain development. Our goal was to expose pregnant rats to propylthiouracil (PTU) to measure the effects of
Si Wu et al.
Journal of pharmaceutical and biomedical analysis, 72, 109-114 (2012-11-14)
A serum metabonomic profiling method based on ultra-performance liquid chromatography/time-of-flight mass spectrometry (UHPLC/TOF-MS) was applied to investigate the metabolic changes in hypothyroid rats induced by propylthiouracil (PTU). With Significance Analysis of Microarray (SAM) for classification and selection of biomarkers, 13
Juneo F Silva et al.
Reproduction, fertility, and development, 24(7), 923-931 (2012-09-01)
The objective of this study was to evaluate fetal weight, histomorphometric changes and proliferative activity, apoptosis and angiogenesis of the placenta in rats with hypothyroidism. Thirty-six adult female rats were divided into two groups with 18 animals each: control and
S A Degrelle et al.
Placenta, 34(1), 29-34 (2012-11-24)
The placenta plays an essential role in the fetomaternal exchanges of iodine and thyroid hormones. Propylthiouracil (PTU) is presently considered to be the treatment of choice for hyperthyroidism during the first trimester of pregnancy. Little is known on the expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service