Skip to Content
Merck
All Photos(4)

Key Documents

C5134

Sigma-Aldrich

2-Chloroadenosine

Synonym(s):

2-CADO, 6-Amino-2-chloropurine riboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClN5O4
CAS Number:
Molecular Weight:
301.69
Beilstein:
43957
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

powder

Quality Level

SMILES string

Nc1nc(Cl)nc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C10H12ClN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6-,9-/m1/s1

InChI key

BIXYYZIIJIXVFW-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloroadenosine has been used:
  • as an agonist of adenosine receptors (A1R) in dorsal hippocampus (DH) and ventral hippocampus (VH) of mice[1]
  • as a component of artificial cerebrospinal fluid (aCSF) for electrophysiological experiments using hippocampal slices[2]
  • as an internal standard in solid phase extraction coupled with liquid chromatography-tandem mass spectrometry (XLC-MS/MS) for the quantification of certain metabolites from axonal lysates and dorsal root ganglia (DRG) neurons[3]

Biochem/physiol Actions

2-Chloroadenosine is an analog of adenosine.[4] It is an adenosine A1 receptor agonist.[5] It might possess anti-oxidant property.[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Sheila Weitzman et al.
Pediatric blood & cancer, 53(7), 1271-1276 (2009-09-05)
A prospective phase II Histiocyte Society study, LCH-S-98, evaluated the efficacy of 2-chlorodeoxyadenosine (2-CdA) monotherapy as salvage therapy in Langerhans cell histiocytosis (LCH). Patients with poor and intermediate risk LCH not responsive to initial therapy and patients with low-risk chronic
2-Chloroadenosine decreases long-term potentiation in the hippocampal CA1 area of the rat
de Mendoncca A and Ribeiro J A
Neuroscience Letters, 118(1), 107-111 (1990)
Effect of locally infused 2-chloroadenosine, an A1 receptor agonist, on spontaneous and evoked dopamine release in rat neostriatum
Ballarin M, et al.
Neuroscience Letters, 185(1), 29-32 (1995)
Wen-Li Gao et al.
Bioresource technology, 118, 82-88 (2012-06-19)
2-Methyltetrahydrofuran (MeTHF), a biomass-derived compound, is a promising medium for biocatalysis and organometallic reactions. The regioselective acylation of 8-chloroadenosine (8-Cl-Ado) and its analogs was carried out in MeTHF with immobilized Penicillium expansum lipase. The lipase displayed more than twofold higher
Javier Díaz-Alonso et al.
eLife, 9 (2020-08-25)
We tested the proposal that the C-terminal domain (CTD) of the AMPAR subunit GluA1 is required for LTP. We found that a knock-in mouse lacking the CTD of GluA1 expresses normal LTP and spatial memory, assayed by the Morris water

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service