Skip to Content
Merck
All Photos(2)

Key Documents

C101

Sigma-Aldrich

8-Cyclopentyl-1,3-dipropylxanthine

solid

Synonym(s):

1,3-Dipropyl-8-cyclopentylxanthine, DPCPX, PD 116,948

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N4O2
CAS Number:
Molecular Weight:
304.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white

solubility

DMSO: >10 mg/mL
0.1 M NaOH: 2 mg/mL
ethanol: 4 mg/mL
H2O: insoluble

εmax

14,800 at 276 nm in ethanol

SMILES string

CCCN1C(=O)N(CCC)c2nc([nH]c2C1=O)C3CCCC3

InChI

1S/C16H24N4O2/c1-3-9-19-14-12(15(21)20(10-4-2)16(19)22)17-13(18-14)11-7-5-6-8-11/h11H,3-10H2,1-2H3,(H,17,18)

InChI key

FFBDFADSZUINTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

8-Cyclopentyl-1,3-dipropylxanthine has been used as an adenosine receptor (A1AR) antagonist in macrophages and human umbilical vein endothelial cells (HUVECs). It has also been used as A1AR antagonist in MCF-7 breast cancer cell line to test its anti-cancer effect.

Biochem/physiol Actions

8-Cyclopentyl-1,3-dipropylxanthine (DPCPX) is a selective A1 adenosine receptor antagonist. DPCPX possesses anti-cancer functionality. It induces apoptosis in breast cancer cells and favors mRNA expression of caspases.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lindsay Silva et al.
Neurotoxicology and teratology, 58, 88-100 (2016-02-24)
Adolescents who use marijuana are more likely to exhibit anxiety, depression, and other mood disorders, including psychotic-like symptoms. Additionally, the age at onset of use and the stress history of the individual can affect responses to cannabis. To examine the
Vanessa R Haynes et al.
Molecular metabolism, 34, 54-71 (2020-03-18)
Nutrient sensing by hypothalamic neurons is critical for the regulation of food intake and energy expenditure. We aimed to identify long- and medium-chain fatty acid species transported into the brain, their effects on energy balance, and the mechanisms by which
Mehdi Nikbakht Dastjerdi et al.
Research in pharmaceutical sciences, 11(4), 303-310 (2016-09-22)
Adenosine receptor family especially A1 type is expressed in breast cancer cells in which P53 and caspase genes are wild-type. The aim of this study was to investigate the correlation between A1 receptor and either cell apoptosis or proliferation and
Hongjie Chen et al.
Experimental hematology, 37(5), 533-538 (2009-04-21)
The control of expression of tumor necrosis factor-alpha (TNF-alpha) impacts a variety of processes during a stress response. Macrophages are a major source of TNF-alpha, the level of which is known to be regulated by adenosine. Previous studies highlighted the
Jordi Bonaventura et al.
Nature communications, 10(1), 4627-4627 (2019-10-13)
Designer Receptors Exclusively Activated by Designer Drugs (DREADDs) are a popular chemogenetic technology for manipulation of neuronal activity in uninstrumented awake animals with potential for human applications as well. The prototypical DREADD agonist clozapine N-oxide (CNO) lacks brain entry and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service