Skip to Content
Merck
Sodium hexanoate 99-100%

Sodium hexanoate

Sodium hexanoate 99-100%
Synonym(s):
Caproic acid sodium salt, Hexanoic acid sodium salt, Sodium caproate
Linear Formula:
CH3(CH2)4COONa
CAS Number:
Molecular Weight:
138.14
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing