Skip to Content
Merck
All Photos(2)

Key Documents

C40

Sigma-Aldrich

Catalase from bovine liver

lyophilized powder, ≥10,000 units/mg protein

Synonym(s):

H2O2:H2O2 oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

form

lyophilized powder

specific activity

≥10,000 units/mg protein

mol wt

tetramer ~250 kDa

contains

≤0.2 wt. % Thymol

isoelectric point

5.4

solubility

50 mM potassium phosphate buffer, pH 7.0: soluble 2.0 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

SMILES string

O(CC)C(=O)c1ccc(cc1)O

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

Gene Information

cow ... CAT(280743)

Looking for similar products? Visit Product Comparison Guide

General description

Research Area: Cell Signaling

Catalase from bovine liver is a tetramer consisting of 4 equal subunits each with a 60 kDa molecular weight. Each of these subunits contains iron bound to a protoheme IX group. The enzyme will also strongly bind to NADP, where NADP and the heme group are within 13.7 angstroms.

Application

Catalase acts as a natural antioxidant to study the roles of reactive oxygen species in gene expression and apoptosis. It has also been used to protect against oxidative damage to proteins, lipids, and nucleic acids. Industrially, catalzes have been used to remove hydrogen peroxide added to milk and cheese, in textile bleaching, and to examine its positive effects on the viability of DNA-repair mutants of E. coli.

Catalase from bovine liver may be used:

  • to prepare H2O2-O2 based biocathode for applications in glucose biofuel cells
  • to study the kinetic properties and storage stability of catalase immobilized on to florisil
  • in glutathione-mediated superoxide generation in an aqueous solution

Biochem/physiol Actions

Catalase activity is constant over the pH range of 4.0-8.5. The enzyme activity is inhibited by 3-amino-1-H-1,2,4 triazole, cyanide, azide, hydroxylamine, cyanogen bromide, 2-mercaptoethanol, dithiothreitol, dianisidine, and nitrate. Incubation of catalase with ascorbate or ascorbate/Cu2+ results in degradation of the catalase molecule.
Catalase, an antioxidant enzyme found in all aerobic organisms, catalyzes the degradation of hydrogen peroxide, a byproduct of metabolic processes, into less harmful water and oxygen. It can also react with alkylhydrogen peroxides, such as methylperoxide and ethylperoxide and the second H2O2 molecule can be replaced by methanol, ethanol, propanol, formate and nitrate as a hydrogen donor. Catalase enzyme uses either iron (Fe) or manganese (Mn) as cofactor, and are classified as Fe-CAT or Mn-CAT.

Caution

Solutions of catalse should not be frozen. Frozen solution will result in a 50-70% loss of activity.

Unit Definition

One unit will decompose 1.0 μmole of H2O2 per min at pH 7.0 at 25 °C, while the H2O2 concentration falls from 10.3 to 9.2 mM, measured by the rate of decrease of A240.

Other Notes

The protein content is determined by the Biuret method. In the product specifications, a protein content >= 70% is guaranteed. The impurities are not analyzed. The only other content specification is the concentration of Thymol, <= 0.2% wt.

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yifan Jiang et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-12)
Clematis florida is a popular ornamental vine species known for diverse colors and shapes of its flowers but not for scent. Here we investigated the composition and biosynthesis of floral scent in 'Kaiser', a fragrant cultivar of C. florida that
Fujun Luo et al.
Nature communications, 12(1), 2380-2380 (2021-04-24)
Diverse signaling complexes are precisely assembled at the presynaptic active zone for dynamic modulation of synaptic transmission and synaptic plasticity. Presynaptic GABAB-receptors nucleate critical signaling complexes regulating neurotransmitter release at most synapses. However, the molecular mechanisms underlying assembly of GABAB-receptor
Afra Khiralla et al.
Microorganisms, 8(9) (2020-09-10)
An endophytic fungus isolated from Vernonia amygdalina, a medicinal plant from Sudan, was taxonomically characterized as Curvularia papendorfii. Ethyl acetate crude extract of C. papendorfii revealed an important antiviral effect against two viral pathogens, the human coronavirus HCoV 229E and
Florian Stehr et al.
Nature communications, 12(1), 4432-4432 (2021-07-23)
Monitoring biomolecules in single-particle tracking experiments is typically achieved by employing fixed organic dyes or fluorescent fusion proteins linked to a target of interest. However, photobleaching typically limits observation times to merely a few seconds, restricting downstream statistical analysis and
Saskia F Erttmann et al.
Nature communications, 10(1), 3493-3493 (2019-08-04)
Hydrogen peroxide (H2O2) has a major function in host-microbial interactions. Although most studies have focused on the endogenous H2O2 produced by immune cells to kill microbes, bacteria can also produce H2O2. How microbial H2O2 influences the dynamics of host-microbial interactions

Protocols

This procedure may be used for all Catalase products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service