Skip to Content
Merck
All Photos(1)

Key Documents

C30

Sigma-Aldrich

Catalase from bovine liver

aqueous suspension, 10,000-40,000 units/mg protein

Synonym(s):

Bovine hepatocatalase, H2O2:H2O2 oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

biological source

bovine liver

form

aqueous suspension

specific activity

10,000-40,000 units/mg protein

mol wt

tetramer ~250 kDa

concentration

20-50 mg/mL

technique(s)

GC/MS: suitable

isoelectric point

5.4

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O(CC)C(=O)c1ccc(cc1)O

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

Gene Information

cow ... CAT(280743)

Looking for similar products? Visit Product Comparison Guide

General description

Catalase from bovine liver contains 506 residues. It is a tetramer and each monomer corresponds to a molecular weight of 61 kDa. The active site in each momomer comprises nicotinamide adenine dinucleotide phosphate (NADPH) and iron binding region.

Research area: Cell Signaling
Research Area: Cell Signaling

Catalase from bovine liver is a tetramer consisting of 4 equal subunits each with a 60 kDa molecular weight. Each of these subunits contains iron bound to a protoheme IX group. The enzyme will also strongly bind to NADP, where NADP and the heme group are within 13.7 angstroms.

Application

Catalase acts as a natural antioxidant to study the roles of reactive oxygen species in gene expression and apoptosis. It has also been used to protect against oxidative damage to proteins, lipids, and nucleic acids. Industrially, catalzes have been used to remove hydrogen peroxide added to milk and cheese, in textile bleaching, and to examine its positive effects on the viability of DNA-repair mutants of E. coli.

Catalase from bovine liver may be used:

  • to prepare H2O2-O2 based biocathode for applications in glucose biofuel cells
  • to study the kinetic properties and storage stability of catalase immobilized on to florisil
  • in glutathione-mediated superoxide generation in an aqueous solution
Catalase from bovine liver has been used for measuring the hydrogen peroxide content in cancer tissue homogenates. It has also been used to test the effect of organophosphate insecticide chlorpyrifos-ethyl (CE) [0,0-diethyl 0 (3,5,6-trichloro-2-pyridyl) phosphorothioate] on its enzyme activity.
Catalase from bovine liver is used for the following applications:
  • Dielectrophoretic field-flow fractionation (DEP-FFF): It is a chromatographic method in which cell elution times reflect the positions of cells in a hydrodynamic flow profile under the control of sedimentation, DEP and hydrodynamic lift forces, FSED, FDEP and FHDL, respectively. The DEP-FFF buffer consists of catalase along with other ingrediants.
  • GC-MS analyses.
  • Deionization.
  • GC-FID analysis.5
  • Functional analysis of blood vessel.

Biochem/physiol Actions

Catalase, an antioxidant enzyme found in all aerobic organisms, catalyzes the degradation of hydrogen peroxide, a byproduct of metabolic processes, into less harmful water and oxygen. It can also react with alkylhydrogen peroxides, such as methylperoxide and ethylperoxide and the second H2O2 molecule can be replaced by methanol, ethanol, propanol, formate and nitrate as a hydrogen donor. Catalase enzyme uses either iron (Fe) or manganese (Mn) as cofactor, and are classified as Fe-CAT or Mn-CAT.
This product doesn′t need any activators, but it is inhibited by 3-amino-1-H-1,2,4 triazole, cyanide, azide, hydroxylamine, cyanogens bromide, 2-mercaptoethanol, dithiothreitol, dianisidnie and nitrate.

Caution

Solutions of catalse should not be frozen. Frozen solution will result in a 50-70% loss of activity.

Unit Definition

One unit will decompose 1.0 μmole of H2O2 per min at pH 7.0 at 25 °C, while the H2O2 concentration falls from 10.3 to 9.2 mM, measured by the rate of decrease of A240.

Preparation Note

This product is a crystalline suspension in water containing 0.1% thymol with activity of 10,000-40,000 units/mg.

Storage and Stability

Tightly closed. Dry. Keep locked up or in an area accessible only to qualified or authorized
persons

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sixty years from discovery to solution: crystal structure of bovine liver catalase form III
Foroughi LM, et al.
Acta Crystallographica Section D, Biological Crystallography, 67(9), 756-762 (2011)
Stéphanie Heuskin et al.
Journal of chromatography. A, 1216(14), 2768-2775 (2008-10-28)
The chemical composition of Matricaria chamomilla L. and Nepeta cataria L. essential oils was determined by GC-MS on an apolar stationary phase by comparison of the characteristic fragmentation patterns with those of the Wiley 275L database. The GC-MS chromatograms were
Sangjo Shim et al.
Biomicrofluidics, 7(1), 11808-11808 (2014-01-10)
The number of circulating tumor cells (CTCs) found in blood is known to be a prognostic marker for recurrence of primary tumors, however, most current methods for isolating CTCs rely on cell surface markers that are not universally expressed by
The complete amino acid sequence of bovine liver catalase and the partial sequence of bovine erythrocyte catalase
Schroeder WA, et al.
Archives of Biochemistry and Biophysics, 214(1), 397-421 (1982)
The effect of organophosphate insecticide chlorpyrifos-ethyl on lipid peroxidation and antioxidant enzymes (in vitro)
Gultekin F, et al.
Archives of Toxicology, 74(9), 533-538 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service