Skip to Content
Merck
All Photos(1)

Key Documents

C3640

Sigma-Aldrich

Carbonic Anhydrase Isozyme II from bovine erythrocytes

lyophilized powder, ≥2,000 W-A units/mg protein

Synonym(s):

Carbonate Dehydratase, Carbonate Hydrolyase, Carbonic Anhydrase II

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

specific activity

≥2,000 W-A units/mg protein

pI 

~5.4

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Carbonic anhydrase from sigma has been used for the analysis of thermodynamic stability of the enzyme.[1] The enzyme has also been used to generate CD4+ T cell lines specific for carbonic anhydrase during the study of autoimmune pancreatitis in rat.[2]

Biochem/physiol Actions

Carbonic anhydrase is a zinc metalloenzyme that has a molecular weight of approximately 30,000 Da. The enzyme catalyzes the hydration of carbon dioxide to carbonic acid. It is involved in vital physiological and pathological processes such as pH and CO2 homeostasis, transport of bicarbonate and CO2, biosynthetic reactions, bone resorption, calcification, and tumorigenicity. Therefore, this enzyme is an important target for inhibitors with clinical applications in pathologies such as glaucoma, epilepsy and Parkinson′s disease.[1]

Unit Definition

One Wilbur-Anderson (W-A) unit will cause the pH of a 0.02 M Trizma buffer to drop from 8.3 to 6.3 per min at 0 °C. (One W-A unit is essentially equivalent to one Roughton-Booth unit.)

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd S Davidson et al.
The American journal of pathology, 166(3), 729-736 (2005-03-04)
Autoimmune pancreatitis (AIP), a recently defined disease of unknown etiology, is characterized by inflammatory infiltrates in the pancreas with conspicuous involvement of the ducts. The disease clinically manifests in humans as epigastric pain, weight loss, and jaundice. This report describes
Daumantas Matulis et al.
Biochemistry, 44(13), 5258-5266 (2005-03-30)
ThermoFluor (a miniaturized high-throughput protein stability assay) was used to analyze the linkage between protein thermal stability and ligand binding. Equilibrium binding ligands increase protein thermal stability by an amount proportional to the concentration and affinity of the ligand. Binding
Frédéric D Birkhäuser et al.
Journal of immunotherapy (Hagerstown, Md. : 1997), 36(2), 102-111 (2013-02-05)
The dendritic cell vaccine DC-Ad-GM·CAIX is an active, specific immunotherapy with the potential of providing a safe and effective therapy against renal cell carcinoma (RCC). Using immunocompetent Balb/c mouse models we tested the efficacy and mechanism of the vaccine to
Dina Schütze et al.
Journal of cancer research and clinical oncology, 139(5), 747-754 (2013-01-30)
Carbonic anhydrase IX (CAIX) is involved in pH homeostasis, growth and survival of tumor cells. Besides the membranous form of CAIX, a soluble form is detectable in serum (s-CAIX). Overexpression of CAIX in tumors offers the opportunity for therapeutic strategies
Xiuhong Shan et al.
Journal of computer assisted tomography, 37(1), 22-28 (2013-01-17)
The objective of this study was to investigate the correlation between the degree of necrosis displayed in computed tomography (CT) image and the expression of hypoxic and angiogenesis biomarkers of breast cancer. Forty-four breast cancer cases were examined with CT

Protocols

Objective: To standardize a procedure for the enzymatic assay of Carbonic Anhydrase (EC 4.2.1.1) for Wilbur-Anderson Units.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service