Skip to Content
Merck
All Photos(2)

Documents

C2624

Sigma-Aldrich

Carbonic Anhydrase from bovine erythrocytes

≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder

Synonym(s):

Carbonic Anhydrase from bovine erythrocytes, Carbonate Dehydratase, Carbonate Hydrolyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine erythrocytes

Assay

≥75% protein basis (biuret)
≥95% (SDS-PAGE)

form

lyophilized powder

specific activity

≥3,500 W-A units/mg protein

solubility

deionized water: >10 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Carbonic anhydrase, from bovine erythrocytes, is used to create carbon dioxide capture systems and to research various purification techniques . Carbonic anhydrase is also used to study acid-base regulation in fish and carbonic anhydrase type II deficiency syndrome .

Biochem/physiol Actions

Carbonic Anhydrase is a zinc-containing enzyme that catalyzes the reversible conversion of carbon dioxide to bicarbonate. One of its main physiological roles is to maintain the acid-base balance in blood and other tissues. Lack of carbonic anhydrase results in carbonic anhydrase type II deficiency syndrome, which is an autosomal recessive disease that causes osteopetrosis, renal tubular acidosis and brain calcifications .

Unit Definition

One Wilbur-Anderson (W-A) unit will cause the pH of a 0.02 M Trizma buffer to drop from 8.3 to 6.3 per min at 0°C. (One W-A unit is essentially equivalent to one Roughton-Booth unit.)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Ghéczy et al.
RSC advances, 10(32), 18655-18676 (2020-05-18)
Enzyme-catalysed cascade reactions in flow-through systems with immobilised enzymes currently are of great interest for exploring their potential for biosynthetic and bioanalytical applications. Basic studies in this field often aim at understanding the stability of the immobilised enzymes and their
Abdur Rauf et al.
Chemistry & biodiversity, 14(6) (2017-02-17)
Plant phenolics are known to display many pharmacological activities. In the current study, eight phenolic compounds, e.g., luteolin 5-O-β-glucoside (1), methyl rosmarinate (2), apigenin (3), vicenin 2 (4), lithospermic acid (5), soyasaponin II (6), rubiadin 3-O-β-primeveroside (7), and 4-(β-d-glucopyranosyloxy)benzyl 3,4-dihydroxybenzoate
Marco Catalano et al.
Analytical chemistry, 92(15), 10822-10829 (2020-07-04)
The availability of reliable methods for the characterization of the binding of small molecule ligands to protein targets is crucially important for drug discovery. We have adapted a method, routinely used for the characterization of monoclonal antibodies (enzyme-linked immunosorbent assay
K M Gilmour et al.
The Journal of experimental biology, 212(Pt 11), 1647-1661 (2009-05-19)
Carbonic anhydrase (CA) is the zinc metalloenzyme that catalyses the reversible reactions of CO(2) with water. CA plays a crucial role in systemic acid-base regulation in fish by providing acid-base equivalents for exchange with the environment. Unlike air-breathing vertebrates, which
Thomas M Bosley et al.
Brain : a journal of neurology, 134(Pt 12), 3502-3515 (2011-11-29)
Carbonic anhydrase type II deficiency syndrome is an uncommon autosomal recessive disease with cardinal features including osteopetrosis, renal tubular acidosis and brain calcifications. We describe the neurological, neuro-ophthalmological and neuroradiological features of 23 individuals (10 males, 13 females; ages at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service