Skip to Content
Merck
All Photos(1)

Key Documents

C174

Sigma-Aldrich

Corticosterone: HBC complex

≥91.5% (HPLC), free flowing powder, water-soluble stress hormone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

Corticosterone: HBC complex, free flowing powder

biological source

synthetic (organic)

sterility

non-sterile

Assay

≥91.5% (HPLC)

form

free flowing powder

shipped in

ambient

storage temp.

room temp

SMILES string

O[C@@H]1[C@H]2[C@H]([C@H]4[C@@]([C@H](CC4)C(=O)CO)(C1)C)CCC3=CC(=O)CC[C@@]32C

InChI

1S/C21H30O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h9,14-17,19,22,24H,3-8,10-11H2,1-2H3/t14-,15-,16+,17-,19+,20-,21-/m0/s1

InChI key

OMFXVFTZEKFJBZ-HJTSIMOOSA-N

Application

Corticosterone: HBC complex has been used:
  • in the intravenous and intraperitoneal administration of corticosterone to rats to test its effect on glucocorticoid and mineralocorticoid activity
  • to induce acute stress in mice
  • in Pulsatile treatment in mice to test its effect on clock gene period 1 transcription

Biochem/physiol Actions

The complex of corticosterone with 2-hydroxypropyl-β-cyclodextrin (HBC) improves water solubility. HBC is a carrier molecule. Corticosterone is a rodent-specific primary adrenal corticosteroid. It displays affinity towards the glucocorticoid and mineralocorticoid receptors.

Reconstitution

Corticosterone content: 66.7 mg/g. When 1 g of complex is reconstituted with water to a total volume of 2.12 mL, a solution which is 90.8 mM in corticosterone is obtained. Corticosterone itself is virtually insoluble in water.

Other Notes

Preformed water-soluble complex of corticosterone and 2-hydroxypropyl-β-cyclodextrin. Package size is total weight of both the corticosterone and the 2-hydroxypropyl-β-cyclodextrin. Corticosterone content: 66.7 mg/g

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marlena Wosiski-Kuhn et al.
Psychoneuroendocrinology, 42, 165-177 (2014-03-19)
Diabetes and obesity are associated with perturbation of adrenal steroid hormones and impairment of hippocampal plasticity, but the question of whether these conditions recruit glucocorticoid-mediated molecular cascades that are comparable to other stressors has yet to be fully addressed. We
Jan Kroon et al.
Molecular metabolism, 47, 101179-101179 (2021-02-07)
Brown adipose tissue (BAT) displays a strong circadian rhythm in metabolic activity, but it is unclear how this rhythm is regulated. As circulating levels of corticosterone coincide with the rhythm of triglyceride-derived fatty acid (FA) uptake by BAT, we investigated
CORT, Cort, B, Corticosterone, and now Cortistatin: Enough Already!
Hershel Raff
Endocrinology, 157(9), 3307-3308 (2016-07-29)
Angela M Ives et al.
Journal of virology, 91(13) (2017-04-14)
Herpes simplex viruses 1 and 2 (HSV-1 and HSV-2) infect and establish latency in peripheral neurons, from which they can reactivate to cause recurrent disease throughout the life of the host. Stress is associated with the exacerbation of clinical symptoms
B L Conway-Campbell et al.
Journal of neuroendocrinology, 22(10), 1093-1100 (2010-07-24)
In vivo glucocorticoid (GC) secretion exhibits a distinctive ultradian rhythmicity. The lipophilic hormone can rapidly diffuse into cells, although only the pulse peak is of sufficient amplitude to activate the low affinity glucocorticoid receptor (GR). Discrete pulses readily access brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service