Skip to Content
Merck
All Photos(4)

Key Documents

C1131

Sigma-Aldrich

Cytidine 5′-monophosphate

Sigma Grade, ≥99% (HPLC), synthetic, powder

Synonym(s):

5′-CMP, 5′-Cytidylic acid, C-5′-P

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N3O8P
CAS Number:
Molecular Weight:
323.20
Beilstein:
46982
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Sigma Grade

Assay

≥99% (HPLC)

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]2O

InChI

1S/C9H14N3O8P/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18)/t4-,6-,7-,8-/m1/s1

InChI key

IERHLVCPSMICTF-XVFCMESISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cytidine 5′-monophosphate has been used in Raman spectroscopy studies.[1]

Biochem/physiol Actions

Cytidine 5′-monophosphate (CMP) is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form CDP which upon phosphorylation to CTP supports DNA and RNA biosynthesis.
The nucleoside diphosphates cytidine diphosphate (CDP) and uridine diphosphate (UDP) are essential for pyrimidine synthesis[2] Cytidine 5′-monophosphate (CMP) nucleotide is useful in adsorption studies on biomimetic apatite for mimicking bone mineral.[3] It also serves an immunity supplement in pediatric formulas.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda J Reha-Krantz et al.
Biochemistry, 50(46), 10136-10149 (2011-10-26)
The adenine base analogue 2-aminopurine (2AP) is a potent base substitution mutagen in prokaryotes because of its enhanceed ability to form a mutagenic base pair with an incoming dCTP. Despite more than 50 years of research, the structure of the
A Hirono et al.
Clinica chimica acta; international journal of clinical chemistry, 130(2), 189-197 (1983-05-30)
Three new variants of pyrimidine 5'-nucleotidase (P5N) found in Japan were studied. They are characterized by slow electrophoretic mobility and a high Michaelis constant for cytidine 5'-monophosphate as has been described in previously reported cases, but are unique with respect
Otto Geiger et al.
Biochimica et biophysica acta, 1831(3), 503-513 (2012-08-28)
Phosphatidylcholine (PC) is the major membrane-forming phospholipid in eukaryotes and is estimated to be present in about 15% of the domain Bacteria. Usually, PC can be synthesized in bacteria by either of two pathways, the phospholipid N-methylation (Pmt) pathway or
Páraic M Keane et al.
Physical chemistry chemical physics : PCCP, 14(18), 6307-6311 (2012-02-24)
The decay pathways of UV-excited cytosine polymers are investigated using picosecond time-resolved infrared spectroscopy. Similar yields of a non-emissive (1)nπ* state are found in the single-stranded dC(30) polymer as in the dCMP monomer, but with a longer lifetime in the
An enzymatic alternative for the synthesis of nucleoside 5?-monophosphates
Gudino ED, et al.
Enzyme and Microbial Technology, 111, 1-6 (2018)

Protocols

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

ZIC®-cHILIC is a densely bonded zwitterionic stationary phase with phosphorylcholine functional groups covalently attached to silica.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service